Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17605953-17606184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM8A1
Entrez Gene ID
51439
Gene Name
family with sequence similarity 8 member A1
Gene Aliases
AHCP
UniGene
Hs.95260
Gene Chromosome Location
Chr.6: 17600287-17611719 on Build GRCh38
Transcripts
NM_016255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660318 c.1050A>G p.A350A Chr.6: 17605966-17605966 on Build GRCh38
4593681 c.1037C>T p.S346L Chr.6: 17605953-17605953 on Build GRCh38
7310111 c.1097+9T>G p.? Chr.6: 17606022-17606022 on Build GRCh38
6339094 c.1052C>T p.P351L Chr.6: 17605968-17605968 on Build GRCh38
4595157 c.1071T>G p.I357M Chr.6: 17605987-17605987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370429714 Chr.6: 17606039-17606039 on Build GRCh38
rs770423837 Chr.6: 17605982-17605982 on Build GRCh38
rs543726619 Chr.6: 17606093-17606093 on Build GRCh38
rs773619983 Chr.6: 17606034-17606035 on Build GRCh38
rs769724371 Chr.6: 17605990-17605990 on Build GRCh38
rs760948246 Chr.6: 17606047-17606047 on Build GRCh38
rs1006799922 Chr.6: 17605980-17605980 on Build GRCh38
rs981541169 Chr.6: 17606148-17606148 on Build GRCh38
rs1049819780 Chr.6: 17606020-17606020 on Build GRCh38
rs144228602 Chr.6: 17606013-17606013 on Build GRCh38
rs75446300 Chr.6: 17605987-17605987 on Build GRCh38
rs989867312 Chr.6: 17606060-17606060 on Build GRCh38
rs79642714 Chr.6: 17605974-17605974 on Build GRCh38
rs780737335 Chr.6: 17605986-17605986 on Build GRCh38
rs772376682 Chr.6: 17606022-17606022 on Build GRCh38
rs201207827 Chr.6: 17606108-17606111 on Build GRCh38
rs1053133152 Chr.6: 17606081-17606081 on Build GRCh38
rs762810325 Chr.6: 17606014-17606014 on Build GRCh38
rs748593283 Chr.6: 17606114-17606118 on Build GRCh38
rs879611214 Chr.6: 17606083-17606083 on Build GRCh38
rs754237033 Chr.6: 17606062-17606062 on Build GRCh38
rs376066332 Chr.6: 17606042-17606042 on Build GRCh38
rs750626887 Chr.6: 17606046-17606046 on Build GRCh38
rs372115838 Chr.6: 17606064-17606064 on Build GRCh38
rs774296980 Chr.6: 17606030-17606030 on Build GRCh38
rs967133806 Chr.6: 17606135-17606135 on Build GRCh38
rs574663175 Chr.6: 17606026-17606026 on Build GRCh38
rs748343756 Chr.6: 17606014-17606015 on Build GRCh38

View Full Product Details