Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144537598-144537859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP39
Entrez Gene ID
80728
Gene Name
Rho GTPase activating protein 39
Gene Aliases
CrGAP, Vilse
UniGene
Hs.148988
Gene Chromosome Location
Chr.8: 144529179-144700520 on Build GRCh38
Transcripts
NM_001308207.1 NM_001308208.1 NM_025251.2 XM_011517308.1 XM_011517309.1 XM_011517312.2 XM_017013870.1 XM_017013871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097300 c.2542C>A p.L848I Chr.8: 144537793-144537793 on Build GRCh38
7086418 c.2602G>C p.E868Q Chr.8: 144537733-144537733 on Build GRCh38
3834307 c.2583G>C p.K861N Chr.8: 144537752-144537752 on Build GRCh38
3646646 c.2558C>T p.T853I Chr.8: 144537777-144537777 on Build GRCh38
3834306 c.2611G>A p.D871N Chr.8: 144537724-144537724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748605530 Chr.8: 144537767-144537767 on Build GRCh38
rs182432238 Chr.8: 144537768-144537768 on Build GRCh38
rs776371042 Chr.8: 144537767-144537769 on Build GRCh38
rs775731470 Chr.8: 144537670-144537670 on Build GRCh38
rs751471405 Chr.8: 144537721-144537721 on Build GRCh38
rs753575968 Chr.8: 144537698-144537698 on Build GRCh38
rs145607062 Chr.8: 144537709-144537709 on Build GRCh38
rs538450932 Chr.8: 144537667-144537667 on Build GRCh38
rs186725620 Chr.8: 144537769-144537769 on Build GRCh38
rs765769497 Chr.8: 144537750-144537765 on Build GRCh38
rs768801452 Chr.8: 144537760-144537760 on Build GRCh38
rs1041309387 Chr.8: 144537661-144537661 on Build GRCh38
rs375961404 Chr.8: 144537809-144537809 on Build GRCh38
rs777818602 Chr.8: 144537716-144537716 on Build GRCh38
rs779000559 Chr.8: 144537763-144537763 on Build GRCh38
rs368831025 Chr.8: 144537818-144537818 on Build GRCh38
rs770586223 Chr.8: 144537787-144537787 on Build GRCh38
rs925714249 Chr.8: 144537624-144537624 on Build GRCh38
rs372291412 Chr.8: 144537800-144537800 on Build GRCh38
rs567900275 Chr.8: 144537633-144537633 on Build GRCh38
rs77443917 Chr.8: 144537706-144537706 on Build GRCh38
rs762230889 Chr.8: 144537688-144537688 on Build GRCh38
rs978433436 Chr.8: 144537623-144537623 on Build GRCh38
rs554174279 Chr.8: 144537705-144537705 on Build GRCh38
rs538932547 Chr.8: 144537693-144537693 on Build GRCh38
rs145613233 Chr.8: 144537726-144537726 on Build GRCh38
rs762853026 Chr.8: 144537738-144537738 on Build GRCh38
rs978850549 Chr.8: 144537839-144537839 on Build GRCh38
rs749526080 Chr.8: 144537742-144537742 on Build GRCh38
rs1022881682 Chr.8: 144537618-144537618 on Build GRCh38
rs749476982 Chr.8: 144537671-144537671 on Build GRCh38
rs752276674 Chr.8: 144537815-144537815 on Build GRCh38
rs368338962 Chr.8: 144537692-144537692 on Build GRCh38
rs1022912583 Chr.8: 144537789-144537789 on Build GRCh38
rs751522875 Chr.8: 144537836-144537836 on Build GRCh38
rs969740923 Chr.8: 144537621-144537621 on Build GRCh38
rs554133355 Chr.8: 144537657-144537657 on Build GRCh38
rs759164654 Chr.8: 144537773-144537773 on Build GRCh38
rs759262457 Chr.8: 144537806-144537806 on Build GRCh38
rs368981280 Chr.8: 144537699-144537699 on Build GRCh38
rs1050306795 Chr.8: 144537691-144537691 on Build GRCh38
rs1053342451 Chr.8: 144537647-144537647 on Build GRCh38
rs201877215 Chr.8: 144537840-144537840 on Build GRCh38
rs944200694 Chr.8: 144537660-144537660 on Build GRCh38
rs914036058 Chr.8: 144537634-144537634 on Build GRCh38
rs934930266 Chr.8: 144537638-144537642 on Build GRCh38
rs112200720 Chr.8: 144537628-144537628 on Build GRCh38
rs199926011 Chr.8: 144537690-144537690 on Build GRCh38
rs769034697 Chr.8: 144537681-144537681 on Build GRCh38
rs556748601 Chr.8: 144537687-144537687 on Build GRCh38

View Full Product Details