Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 195289078-195289351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP2
Entrez Gene ID
23527
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Gene Aliases
CENTB2, CNT-B2
UniGene
Hs.593373
Gene Chromosome Location
Chr.3: 195274736-195443176 on Build GRCh38
Transcripts
NM_012287.5 XM_006713557.3 XM_011512602.2 XM_011512603.2 XM_011512604.2 XM_011512605.2 XM_011512606.2 XM_017006046.1 XM_017006047.1 XM_017006048.1 XM_017006049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042762 c.2064G>A p.G688G Chr.3: 195289231-195289231 on Build GRCh38
5638257 c.2155A>C p.N719H Chr.3: 195289140-195289140 on Build GRCh38
479875 c.2108C>T p.T703I Chr.3: 195289187-195289187 on Build GRCh38
318470 c.2119G>T p.G707W Chr.3: 195289176-195289176 on Build GRCh38
318469 c.2120G>C p.G707A Chr.3: 195289175-195289175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745940118 Chr.3: 195289176-195289176 on Build GRCh38
rs752457653 Chr.3: 195289277-195289277 on Build GRCh38
rs1044393243 Chr.3: 195289113-195289113 on Build GRCh38
rs777983830 Chr.3: 195289130-195289130 on Build GRCh38
rs115363377 Chr.3: 195289265-195289265 on Build GRCh38
rs757495309 Chr.3: 195289146-195289146 on Build GRCh38
rs371627874 Chr.3: 195289218-195289218 on Build GRCh38
rs754114645 Chr.3: 195289139-195289139 on Build GRCh38
rs774216426 Chr.3: 195289263-195289263 on Build GRCh38
rs141544377 Chr.3: 195289291-195289291 on Build GRCh38
rs766227398 Chr.3: 195289261-195289261 on Build GRCh38
rs767365534 Chr.3: 195289266-195289266 on Build GRCh38
rs572459177 Chr.3: 195289286-195289286 on Build GRCh38
rs776647872 Chr.3: 195289282-195289282 on Build GRCh38
rs780570592 Chr.3: 195289199-195289199 on Build GRCh38
rs778900639 Chr.3: 195289172-195289172 on Build GRCh38
rs148271258 Chr.3: 195289234-195289234 on Build GRCh38
rs777124456 Chr.3: 195289226-195289226 on Build GRCh38
rs760470012 Chr.3: 195289270-195289270 on Build GRCh38
rs769462393 Chr.3: 195289241-195289241 on Build GRCh38
rs769089048 Chr.3: 195289225-195289225 on Build GRCh38
rs762481020 Chr.3: 195289253-195289253 on Build GRCh38
rs888367247 Chr.3: 195289231-195289231 on Build GRCh38
rs772617734 Chr.3: 195289243-195289243 on Build GRCh38
rs778689853 Chr.3: 195289271-195289272 on Build GRCh38
rs200221117 Chr.3: 195289305-195289306 on Build GRCh38
rs900202535 Chr.3: 195289267-195289267 on Build GRCh38
rs944651047 Chr.3: 195289167-195289167 on Build GRCh38
rs139073809 Chr.3: 195289184-195289184 on Build GRCh38

View Full Product Details