Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30007032-30007291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2A
Entrez Gene ID
8448
Gene Name
double C2 domain alpha
Gene Aliases
Doc2
UniGene
Hs.355281
Gene Chromosome Location
Chr.16: 30005514-30023280 on Build GRCh38
Transcripts
NM_001282062.1 NM_001282063.1 NM_001282068.1 NM_003586.2 NR_104089.1 NR_104090.1 XM_011545975.1 XM_017023776.1 XM_017023777.1 XM_017023778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1301850 c.640G>A p.E214K Chr.16: 30007187-30007187 on Build GRCh38
3888381 c.576G>A p.G192G Chr.16: 30007251-30007251 on Build GRCh38
3508709 c.672C>T p.S224S Chr.16: 30007073-30007073 on Build GRCh38
3508710 c.577G>A p.E193K Chr.16: 30007250-30007250 on Build GRCh38
4554316 c.618G>A p.K206K Chr.16: 30007209-30007209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200639006 Chr.16: 30007216-30007216 on Build GRCh38
rs753409511 Chr.16: 30007183-30007183 on Build GRCh38
rs750106154 Chr.16: 30007188-30007188 on Build GRCh38
rs764082327 Chr.16: 30007182-30007182 on Build GRCh38
rs781582421 Chr.16: 30007203-30007203 on Build GRCh38
rs753651850 Chr.16: 30007255-30007255 on Build GRCh38
rs945021350 Chr.16: 30007084-30007084 on Build GRCh38
rs758051622 Chr.16: 30007196-30007196 on Build GRCh38
rs765857200 Chr.16: 30007259-30007259 on Build GRCh38
rs202009086 Chr.16: 30007065-30007065 on Build GRCh38
rs376675512 Chr.16: 30007218-30007218 on Build GRCh38
rs147170771 Chr.16: 30007228-30007228 on Build GRCh38
rs752519763 Chr.16: 30007176-30007176 on Build GRCh38
rs762895181 Chr.16: 30007147-30007147 on Build GRCh38
rs767570558 Chr.16: 30007114-30007114 on Build GRCh38
rs538503237 Chr.16: 30007143-30007143 on Build GRCh38
rs577757695 Chr.16: 30007123-30007123 on Build GRCh38
rs756963479 Chr.16: 30007132-30007132 on Build GRCh38
rs778379217 Chr.16: 30007134-30007134 on Build GRCh38
rs765364473 Chr.16: 30007085-30007085 on Build GRCh38
rs749009321 Chr.16: 30007057-30007057 on Build GRCh38
rs773317733 Chr.16: 30007145-30007145 on Build GRCh38
rs759800024 Chr.16: 30007164-30007164 on Build GRCh38
rs767478540 Chr.16: 30007173-30007173 on Build GRCh38
rs775802877 Chr.16: 30007250-30007250 on Build GRCh38
rs373818256 Chr.16: 30007120-30007120 on Build GRCh38
rs756663763 Chr.16: 30007184-30007184 on Build GRCh38
rs770987510 Chr.16: 30007229-30007229 on Build GRCh38
rs906252063 Chr.16: 30007121-30007121 on Build GRCh38
rs140372679 Chr.16: 30007242-30007242 on Build GRCh38
rs751394505 Chr.16: 30007102-30007102 on Build GRCh38
rs747414032 Chr.16: 30007135-30007135 on Build GRCh38
rs764137925 Chr.16: 30007251-30007251 on Build GRCh38
rs769227502 Chr.16: 30007064-30007064 on Build GRCh38
rs778631184 Chr.16: 30007187-30007187 on Build GRCh38
rs1031612341 Chr.16: 30007264-30007264 on Build GRCh38
rs142406537 Chr.16: 30007244-30007244 on Build GRCh38
rs747294200 Chr.16: 30007062-30007062 on Build GRCh38
rs771202623 Chr.16: 30007150-30007150 on Build GRCh38
rs3935873 Chr.16: 30007179-30007179 on Build GRCh38
rs1000516839 Chr.16: 30007096-30007096 on Build GRCh38
rs774373553 Chr.16: 30007160-30007160 on Build GRCh38
rs371218364 Chr.16: 30007136-30007136 on Build GRCh38
rs770468720 Chr.16: 30007060-30007060 on Build GRCh38
rs374821872 Chr.16: 30007243-30007243 on Build GRCh38
rs759726649 Chr.16: 30007236-30007236 on Build GRCh38
rs1051175353 Chr.16: 30007206-30007208 on Build GRCh38
rs191060572 Chr.16: 30007267-30007267 on Build GRCh38
rs367662928 Chr.16: 30007125-30007125 on Build GRCh38
rs907218911 Chr.16: 30007219-30007236 on Build GRCh38
rs557405576 Chr.16: 30007162-30007162 on Build GRCh38
rs766473626 Chr.16: 30007097-30007097 on Build GRCh38
rs34789550 Chr.16: 30007253-30007254 on Build GRCh38
rs761980118 Chr.16: 30007142-30007142 on Build GRCh38
rs139431507 Chr.16: 30007208-30007208 on Build GRCh38
rs748708048 Chr.16: 30007138-30007138 on Build GRCh38
rs370418043 Chr.16: 30007093-30007093 on Build GRCh38
rs761882236 Chr.16: 30007083-30007083 on Build GRCh38
rs143560174 Chr.16: 30007215-30007215 on Build GRCh38
rs773264300 Chr.16: 30007087-30007087 on Build GRCh38
rs754675552 Chr.16: 30007110-30007110 on Build GRCh38
rs552708306 Chr.16: 30007088-30007088 on Build GRCh38
rs534605224 Chr.16: 30007061-30007061 on Build GRCh38
rs774476924 Chr.16: 30007230-30007230 on Build GRCh38

View Full Product Details