Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27627733-27627958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC121
Entrez Gene ID
79635
Gene Name
coiled-coil domain containing 121
Gene Aliases
-
UniGene
Hs.21081
Gene Chromosome Location
Chr.2: 27625638-27629031 on Build GRCh38
Transcripts
NM_001142683.2 NM_024584.4 XM_005264560.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6621382 c.45G>A p.R15R Chr.2: 27627755-27627755 on Build GRCh38
1752516 c.498G>C p.L166L Chr.2: 27627788-27627788 on Build GRCh38
1752517 c.12G>C p.L4L Chr.2: 27627788-27627788 on Build GRCh38
5713056 c.493delG p.D165fs*2 Chr.2: 27627793-27627793 on Build GRCh38
2940595 c.401C>T p.P134L Chr.2: 27627885-27627885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59370311 Chr.2: 27627765-27627766 on Build GRCh38
rs929300531 Chr.2: 27627766-27627766 on Build GRCh38
rs368304327 Chr.2: 27627786-27627786 on Build GRCh38
rs754587918 Chr.2: 27627792-27627792 on Build GRCh38
rs762903530 Chr.2: 27627885-27627885 on Build GRCh38
rs755726431 Chr.2: 27627915-27627915 on Build GRCh38
rs747799292 Chr.2: 27627805-27627805 on Build GRCh38
rs760719725 Chr.2: 27627831-27627831 on Build GRCh38
rs765387584 Chr.2: 27627865-27627865 on Build GRCh38
rs573333820 Chr.2: 27627904-27627904 on Build GRCh38
rs769354652 Chr.2: 27627817-27627817 on Build GRCh38
rs750232416 Chr.2: 27627876-27627876 on Build GRCh38
rs150375740 Chr.2: 27627776-27627776 on Build GRCh38
rs555033711 Chr.2: 27627896-27627896 on Build GRCh38
rs765053048 Chr.2: 27627756-27627756 on Build GRCh38
rs766399612 Chr.2: 27627890-27627890 on Build GRCh38
rs751105017 Chr.2: 27627781-27627781 on Build GRCh38
rs558290334 Chr.2: 27627763-27627763 on Build GRCh38
rs372061100 Chr.2: 27627846-27627846 on Build GRCh38
rs758349067 Chr.2: 27627771-27627771 on Build GRCh38
rs1051719054 Chr.2: 27627785-27627785 on Build GRCh38
rs943491753 Chr.2: 27627804-27627804 on Build GRCh38
rs777354922 Chr.2: 27627921-27627921 on Build GRCh38
rs776033795 Chr.2: 27627830-27627830 on Build GRCh38
rs779280087 Chr.2: 27627819-27627819 on Build GRCh38
rs766308983 Chr.2: 27627923-27627924 on Build GRCh38
rs746236678 Chr.2: 27627824-27627824 on Build GRCh38
rs776602104 Chr.2: 27627844-27627844 on Build GRCh38
rs187352025 Chr.2: 27627834-27627834 on Build GRCh38
rs143529534 Chr.2: 27627793-27627793 on Build GRCh38
rs780680207 Chr.2: 27627794-27627794 on Build GRCh38
rs904715150 Chr.2: 27627925-27627925 on Build GRCh38

View Full Product Details