Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140402093-140402322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKHD1
Entrez Gene ID
54882
Gene Name
ankyrin repeat and KH domain containing 1
Gene Aliases
MASK, MASK1, PP2500, VBARP
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140539856 on Build GRCh38
Transcripts
NM_001197030.1 NM_017747.2 NM_017978.2 NM_024668.3 NM_020690.5

Gene Symbol
ANKHD1-EIF4EBP3
Entrez Gene ID
404734
Gene Name
ANKHD1-EIF4EBP3 readthrough
Gene Aliases
MASK-BP3, MASK-BP3ARF
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140549578 on Build GRCh38
Transcripts
NM_001197030.1 NM_017747.2 NM_017978.2 NM_024668.3 NM_020690.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182794 c.184G>A p.G62S Chr.5: 140402151-140402151 on Build GRCh38
4585448 c.175_176insGCG p.G63_S64insG Chr.5: 140402142-140402143 on Build GRCh38
7355532 c.182G>A p.G61D Chr.5: 140402149-140402149 on Build GRCh38
1182796 c.184G>A p.G62S Chr.5: 140402151-140402151 on Build GRCh38
5884117 c.134G>A p.R45K Chr.5: 140402101-140402101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377350721 Chr.5: 140402287-140402287 on Build GRCh38
rs1048183735 Chr.5: 140402213-140402213 on Build GRCh38
rs781017416 Chr.5: 140402230-140402230 on Build GRCh38
rs756238079 Chr.5: 140402279-140402279 on Build GRCh38
rs778745048 Chr.5: 140402162-140402163 on Build GRCh38
rs369332517 Chr.5: 140402218-140402218 on Build GRCh38
rs752784412 Chr.5: 140402261-140402261 on Build GRCh38
rs373301863 Chr.5: 140402276-140402276 on Build GRCh38
rs770011324 Chr.5: 140402145-140402145 on Build GRCh38
rs1054506401 Chr.5: 140402168-140402168 on Build GRCh38
rs1032360649 Chr.5: 140402122-140402122 on Build GRCh38
rs763677213 Chr.5: 140402139-140402140 on Build GRCh38
rs200859256 Chr.5: 140402177-140402177 on Build GRCh38
rs754392143 Chr.5: 140402136-140402137 on Build GRCh38
rs766158610 Chr.5: 140402207-140402207 on Build GRCh38
rs751122692 Chr.5: 140402143-140402148 on Build GRCh38
rs779877639 Chr.5: 140402152-140402157 on Build GRCh38
rs767139631 Chr.5: 140402143-140402145 on Build GRCh38
rs572858796 Chr.5: 140402258-140402258 on Build GRCh38
rs778108331 Chr.5: 140402149-140402149 on Build GRCh38
rs754945725 Chr.5: 140402154-140402168 on Build GRCh38
rs760180274 Chr.5: 140402137-140402142 on Build GRCh38
rs1042606577 Chr.5: 140402256-140402258 on Build GRCh38
rs762652307 Chr.5: 140402194-140402194 on Build GRCh38
rs781270043 Chr.5: 140402133-140402133 on Build GRCh38
rs368734871 Chr.5: 140402299-140402299 on Build GRCh38
rs905586631 Chr.5: 140402216-140402216 on Build GRCh38
rs539510939 Chr.5: 140402162-140402162 on Build GRCh38
rs756739194 Chr.5: 140402142-140402143 on Build GRCh38
rs749197636 Chr.5: 140402151-140402152 on Build GRCh38
rs779873630 Chr.5: 140402111-140402111 on Build GRCh38
rs754889342 Chr.5: 140402225-140402225 on Build GRCh38
rs374152319 Chr.5: 140402115-140402115 on Build GRCh38
rs540262672 Chr.5: 140402290-140402290 on Build GRCh38
rs748193536 Chr.5: 140402135-140402135 on Build GRCh38
rs776156839 Chr.5: 140402175-140402175 on Build GRCh38
rs771405517 Chr.5: 140402172-140402172 on Build GRCh38
rs755898462 Chr.5: 140402146-140402157 on Build GRCh38
rs766547412 Chr.5: 140402255-140402255 on Build GRCh38
rs751723859 Chr.5: 140402241-140402241 on Build GRCh38
rs930951565 Chr.5: 140402199-140402199 on Build GRCh38
rs557914339 Chr.5: 140402183-140402183 on Build GRCh38
rs750056985 Chr.5: 140402190-140402190 on Build GRCh38
rs953900550 Chr.5: 140402132-140402132 on Build GRCh38
rs749566779 Chr.5: 140402157-140402157 on Build GRCh38
rs761257889 Chr.5: 140402180-140402180 on Build GRCh38
rs918280276 Chr.5: 140402193-140402193 on Build GRCh38
rs774918744 Chr.5: 140402174-140402174 on Build GRCh38
rs754942185 Chr.5: 140402119-140402119 on Build GRCh38
rs751502542 Chr.5: 140402116-140402116 on Build GRCh38

View Full Product Details