Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11650707-11650952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO2
Entrez Gene ID
26232
Gene Name
F-box protein 2
Gene Aliases
FBG1, FBX2, Fbs1, NFB42, OCP1
UniGene
Hs.132753
Gene Chromosome Location
Chr.1: 11648361-11654831 on Build GRCh38
Transcripts
NM_012168.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600611 c.67G>T p.E23* Chr.1: 11650790-11650790 on Build GRCh38
6392013 c.136T>C p.Y46H Chr.1: 11650721-11650721 on Build GRCh38
5496319 c.123G>C p.A41A Chr.1: 11650734-11650734 on Build GRCh38
1744890 c.133_134insCGC p.A45_Y46insP Chr.1: 11650723-11650724 on Build GRCh38
1180447 c.134_135insCGC p.A45_Y46insA Chr.1: 11650722-11650723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890597013 Chr.1: 11650927-11650927 on Build GRCh38
rs1045681810 Chr.1: 11650790-11650790 on Build GRCh38
rs61773907 Chr.1: 11650934-11650934 on Build GRCh38
rs992599787 Chr.1: 11650796-11650796 on Build GRCh38
rs746688553 Chr.1: 11650737-11650737 on Build GRCh38
rs985768341 Chr.1: 11650752-11650752 on Build GRCh38
rs763514760 Chr.1: 11650777-11650777 on Build GRCh38
rs61749273 Chr.1: 11650734-11650734 on Build GRCh38
rs538764329 Chr.1: 11650725-11650726 on Build GRCh38
rs367609409 Chr.1: 11650806-11650806 on Build GRCh38
rs930483787 Chr.1: 11650762-11650762 on Build GRCh38
rs112380988 Chr.1: 11650724-11650724 on Build GRCh38
rs771682251 Chr.1: 11650730-11650730 on Build GRCh38
rs572164329 Chr.1: 11650748-11650748 on Build GRCh38
rs777280598 Chr.1: 11650869-11650869 on Build GRCh38
rs769151344 Chr.1: 11650852-11650853 on Build GRCh38
rs374950803 Chr.1: 11650803-11650803 on Build GRCh38
rs751222805 Chr.1: 11650729-11650730 on Build GRCh38
rs754920696 Chr.1: 11650826-11650826 on Build GRCh38
rs865995820 Chr.1: 11650817-11650817 on Build GRCh38
rs985139461 Chr.1: 11650764-11650764 on Build GRCh38
rs1030604255 Chr.1: 11650925-11650925 on Build GRCh38
rs770139868 Chr.1: 11650741-11650741 on Build GRCh38
rs868820610 Chr.1: 11650807-11650807 on Build GRCh38
rs192416753 Chr.1: 11650903-11650903 on Build GRCh38
rs778450432 Chr.1: 11650754-11650754 on Build GRCh38
rs1012283081 Chr.1: 11650731-11650731 on Build GRCh38
rs746606559 Chr.1: 11650880-11650880 on Build GRCh38
rs983173287 Chr.1: 11650725-11650725 on Build GRCh38
rs996407736 Chr.1: 11650920-11650920 on Build GRCh38
rs775726014 Chr.1: 11650757-11650757 on Build GRCh38
rs766191218 Chr.1: 11650812-11650812 on Build GRCh38
rs893330026 Chr.1: 11650840-11650840 on Build GRCh38
rs927193469 Chr.1: 11650776-11650776 on Build GRCh38
rs762123854 Chr.1: 11650873-11650873 on Build GRCh38
rs902598165 Chr.1: 11650845-11650845 on Build GRCh38
rs934706302 Chr.1: 11650824-11650824 on Build GRCh38
rs745338626 Chr.1: 11650757-11650757 on Build GRCh38
rs560693746 Chr.1: 11650823-11650823 on Build GRCh38
rs549397727 Chr.1: 11650888-11650888 on Build GRCh38
rs915813236 Chr.1: 11650773-11650773 on Build GRCh38
rs919120976 Chr.1: 11650760-11650760 on Build GRCh38
rs953184967 Chr.1: 11650738-11650738 on Build GRCh38
rs971891922 Chr.1: 11650759-11650759 on Build GRCh38
rs778861115 Chr.1: 11650829-11650829 on Build GRCh38
rs1011734299 Chr.1: 11650847-11650847 on Build GRCh38
rs890013641 Chr.1: 11650879-11650879 on Build GRCh38
rs184979619 Chr.1: 11650839-11650839 on Build GRCh38
rs965751021 Chr.1: 11650756-11650756 on Build GRCh38
rs767723833 Chr.1: 11650804-11650804 on Build GRCh38
rs756805231 Chr.1: 11650732-11650734 on Build GRCh38
rs758438947 Chr.1: 11650748-11650749 on Build GRCh38
rs748115503 Chr.1: 11650838-11650838 on Build GRCh38
rs774504936 Chr.1: 11650775-11650775 on Build GRCh38
rs764571902 Chr.1: 11650781-11650781 on Build GRCh38
rs774554868 Chr.1: 11650799-11650799 on Build GRCh38

View Full Product Details