Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34101767-34101992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC7
Entrez Gene ID
56851
Gene Name
ER membrane protein complex subunit 7
Gene Aliases
C11orf3, C15orf24, HT022, ORF1-FL1
UniGene
Hs.160565
Gene Chromosome Location
Chr.15: 34084022-34101852 on Build GRCh38
Transcripts
NM_020154.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7092065 c.53C>T p.S18L Chr.15: 34101787-34101787 on Build GRCh38
331786 c.19G>T p.G7C Chr.15: 34101821-34101821 on Build GRCh38
7114740 c.66G>A p.Q22Q Chr.15: 34101774-34101774 on Build GRCh38
347041 c.4G>C p.A2P Chr.15: 34101836-34101836 on Build GRCh38
1723871 c.48_50delGCT p.L17delL Chr.15: 34101790-34101792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036450091 Chr.15: 34101962-34101962 on Build GRCh38
rs544388848 Chr.15: 34101906-34101906 on Build GRCh38
rs542176202 Chr.15: 34101939-34101939 on Build GRCh38
rs199721583 Chr.15: 34101829-34101829 on Build GRCh38
rs775691710 Chr.15: 34101835-34101835 on Build GRCh38
rs79363435 Chr.15: 34101832-34101832 on Build GRCh38
rs200838781 Chr.15: 34101819-34101819 on Build GRCh38
rs773745213 Chr.15: 34101863-34101863 on Build GRCh38
rs958456931 Chr.15: 34101883-34101883 on Build GRCh38
rs186499996 Chr.15: 34101910-34101910 on Build GRCh38
rs777828275 Chr.15: 34101789-34101790 on Build GRCh38
rs999647646 Chr.15: 34101864-34101864 on Build GRCh38
rs989720987 Chr.15: 34101902-34101902 on Build GRCh38
rs754548142 Chr.15: 34101804-34101804 on Build GRCh38
rs11557326 Chr.15: 34101816-34101816 on Build GRCh38
rs1031130556 Chr.15: 34101876-34101876 on Build GRCh38
rs564575267 Chr.15: 34101909-34101909 on Build GRCh38
rs768764006 Chr.15: 34101841-34101841 on Build GRCh38
rs757022675 Chr.15: 34101825-34101825 on Build GRCh38
rs957844389 Chr.15: 34101938-34101938 on Build GRCh38
rs777438470 Chr.15: 34101812-34101812 on Build GRCh38
rs745799317 Chr.15: 34101831-34101831 on Build GRCh38
rs766819678 Chr.15: 34101869-34101869 on Build GRCh38
rs762816075 Chr.15: 34101955-34101955 on Build GRCh38
rs752576757 Chr.15: 34101848-34101848 on Build GRCh38
rs780646792 Chr.15: 34101810-34101810 on Build GRCh38
rs752232714 Chr.15: 34101879-34101879 on Build GRCh38
rs762140693 Chr.15: 34101846-34101846 on Build GRCh38
rs776854957 Chr.15: 34101844-34101844 on Build GRCh38
rs928025270 Chr.15: 34101952-34101952 on Build GRCh38
rs982137172 Chr.15: 34101930-34101930 on Build GRCh38
rs575053659 Chr.15: 34101914-34101914 on Build GRCh38
rs554533846 Chr.15: 34101795-34101795 on Build GRCh38
rs201752471 Chr.15: 34101811-34101811 on Build GRCh38
rs747037014 Chr.15: 34101840-34101840 on Build GRCh38
rs907331592 Chr.15: 34101895-34101895 on Build GRCh38
rs756436394 Chr.15: 34101790-34101792 on Build GRCh38
rs765629639 Chr.15: 34101859-34101859 on Build GRCh38

View Full Product Details