Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7372884-7373155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL28A1
Entrez Gene ID
340267
Gene Name
collagen type XXVIII alpha 1 chain
Gene Aliases
COL28
UniGene
Hs.491104
Gene Chromosome Location
Chr.7: 7338543-7542062 on Build GRCh38
Transcripts
NM_001037763.2 XM_011515358.2 XM_011515359.2 XM_011515360.2 XM_011515362.2 XM_017012131.1 XM_017012132.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584659 c.2808G>C p.K936N Chr.7: 7373098-7373098 on Build GRCh38
6012617 c.2794A>G p.I932V Chr.7: 7373112-7373112 on Build GRCh38
1623093 c.2908+10G>A p.? Chr.7: 7372988-7372988 on Build GRCh38
3640522 c.2819C>T p.P940L Chr.7: 7373087-7373087 on Build GRCh38
7204936 c.2830A>T p.I944L Chr.7: 7373076-7373076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946276612 Chr.7: 7373114-7373114 on Build GRCh38
rs917277436 Chr.7: 7373103-7373103 on Build GRCh38
rs750836595 Chr.7: 7373116-7373116 on Build GRCh38
rs190160366 Chr.7: 7372941-7372941 on Build GRCh38
rs767546617 Chr.7: 7373055-7373055 on Build GRCh38
rs201791013 Chr.7: 7373025-7373025 on Build GRCh38
rs578148010 Chr.7: 7372950-7372950 on Build GRCh38
rs755650770 Chr.7: 7373118-7373118 on Build GRCh38
rs995776213 Chr.7: 7372985-7372985 on Build GRCh38
rs764577231 Chr.7: 7373104-7373104 on Build GRCh38
rs768054073 Chr.7: 7373113-7373113 on Build GRCh38
rs375730266 Chr.7: 7373014-7373014 on Build GRCh38
rs373691136 Chr.7: 7372970-7372970 on Build GRCh38
rs776752390 Chr.7: 7372968-7372968 on Build GRCh38
rs200467012 Chr.7: 7373024-7373024 on Build GRCh38
rs780888062 Chr.7: 7373006-7373006 on Build GRCh38
rs371977309 Chr.7: 7373026-7373026 on Build GRCh38
rs775786820 Chr.7: 7373089-7373089 on Build GRCh38
rs755780083 Chr.7: 7373016-7373016 on Build GRCh38
rs553624682 Chr.7: 7372945-7372945 on Build GRCh38
rs776644922 Chr.7: 7373080-7373080 on Build GRCh38
rs888470673 Chr.7: 7372936-7372936 on Build GRCh38
rs748129753 Chr.7: 7373051-7373051 on Build GRCh38
rs567843737 Chr.7: 7372932-7372932 on Build GRCh38
rs992812402 Chr.7: 7373086-7373086 on Build GRCh38
rs761650167 Chr.7: 7372997-7372997 on Build GRCh38
rs556671634 Chr.7: 7372965-7372965 on Build GRCh38
rs201574213 Chr.7: 7373028-7373028 on Build GRCh38
rs61743298 Chr.7: 7373132-7373132 on Build GRCh38
rs200786801 Chr.7: 7372974-7372974 on Build GRCh38
rs764487213 Chr.7: 7372988-7372988 on Build GRCh38
rs762476204 Chr.7: 7373107-7373107 on Build GRCh38
rs774273703 Chr.7: 7372952-7372952 on Build GRCh38
rs765763071 Chr.7: 7373120-7373120 on Build GRCh38
rs753546944 Chr.7: 7373129-7373129 on Build GRCh38
rs763229053 Chr.7: 7373098-7373098 on Build GRCh38
rs372795400 Chr.7: 7373005-7373005 on Build GRCh38
rs757791278 Chr.7: 7373004-7373004 on Build GRCh38
rs61399779 Chr.7: 7372989-7372989 on Build GRCh38
rs751939247 Chr.7: 7373106-7373106 on Build GRCh38
rs140427575 Chr.7: 7373085-7373085 on Build GRCh38
rs1029380094 Chr.7: 7373002-7373002 on Build GRCh38
rs769708782 Chr.7: 7373088-7373088 on Build GRCh38
rs778722209 Chr.7: 7373134-7373134 on Build GRCh38
rs778627958 Chr.7: 7373033-7373033 on Build GRCh38
rs771968392 Chr.7: 7373067-7373067 on Build GRCh38
rs545691658 Chr.7: 7372956-7372956 on Build GRCh38
rs562127536 Chr.7: 7372921-7372921 on Build GRCh38

View Full Product Details