Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37188040-37188267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF6
Entrez Gene ID
114904
Gene Name
C1q and tumor necrosis factor related protein 6
Gene Aliases
CTFP6, CTRP6, ZACRP6
UniGene
Hs.22011
Gene Chromosome Location
Chr.22: 37180166-37199400 on Build GRCh38
Transcripts
NM_031910.3 NM_182486.1 XM_011529857.1 XM_017028569.1 XM_017028570.1 XM_017028571.1 XM_017028572.1 XM_017028573.1 XM_017028574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6456642 c.38C>A p.A13D Chr.22: 37188176-37188176 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs942158423 Chr.22: 37188213-37188213 on Build GRCh38
rs112112832 Chr.22: 37188150-37188150 on Build GRCh38
rs751028282 Chr.22: 37188167-37188167 on Build GRCh38
rs763741450 Chr.22: 37188236-37188236 on Build GRCh38
rs371306732 Chr.22: 37188225-37188225 on Build GRCh38
rs756698009 Chr.22: 37188168-37188168 on Build GRCh38
rs983332228 Chr.22: 37188200-37188200 on Build GRCh38
rs765002650 Chr.22: 37188142-37188142 on Build GRCh38
rs747027209 Chr.22: 37188195-37188195 on Build GRCh38
rs773179223 Chr.22: 37188194-37188194 on Build GRCh38
rs910152997 Chr.22: 37188211-37188211 on Build GRCh38
rs571934831 Chr.22: 37188068-37188068 on Build GRCh38
rs751352140 Chr.22: 37188164-37188165 on Build GRCh38
rs777763647 Chr.22: 37188137-37188138 on Build GRCh38
rs763542092 Chr.22: 37188166-37188166 on Build GRCh38
rs151217898 Chr.22: 37188188-37188188 on Build GRCh38
rs374121913 Chr.22: 37188128-37188128 on Build GRCh38
rs985886340 Chr.22: 37188103-37188103 on Build GRCh38
rs1030613106 Chr.22: 37188067-37188067 on Build GRCh38
rs924622920 Chr.22: 37188135-37188135 on Build GRCh38
rs759394570 Chr.22: 37188140-37188140 on Build GRCh38
rs934775947 Chr.22: 37188138-37188138 on Build GRCh38
rs749804810 Chr.22: 37188187-37188187 on Build GRCh38
rs367991314 Chr.22: 37188132-37188132 on Build GRCh38
rs770995358 Chr.22: 37188217-37188217 on Build GRCh38
rs555023444 Chr.22: 37188191-37188191 on Build GRCh38
rs569624873 Chr.22: 37188173-37188173 on Build GRCh38
rs955851538 Chr.22: 37188083-37188083 on Build GRCh38
rs201852793 Chr.22: 37188118-37188118 on Build GRCh38
rs748353059 Chr.22: 37188190-37188190 on Build GRCh38
rs547351782 Chr.22: 37188065-37188065 on Build GRCh38
rs534478364 Chr.22: 37188235-37188235 on Build GRCh38
rs111465350 Chr.22: 37188106-37188106 on Build GRCh38
rs758037428 Chr.22: 37188148-37188148 on Build GRCh38
rs773305734 Chr.22: 37188125-37188125 on Build GRCh38
rs532963679 Chr.22: 37188079-37188079 on Build GRCh38
rs536582265 Chr.22: 37188075-37188075 on Build GRCh38
rs200211481 Chr.22: 37188247-37188247 on Build GRCh38
rs960122349 Chr.22: 37188198-37188198 on Build GRCh38
rs748382583 Chr.22: 37188117-37188117 on Build GRCh38
rs567158819 Chr.22: 37188234-37188234 on Build GRCh38

View Full Product Details