Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28482186-28482411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN3
Entrez Gene ID
1201
Gene Name
CLN3, battenin
Gene Aliases
BTS, JNCL
UniGene
Hs.534667
Gene Chromosome Location
Chr.16: 28466653-28492302 on Build GRCh38
Transcripts
NM_000086.2 NM_001042432.1 NM_001286104.1 NM_001286105.1 NM_001286109.1 NM_001286110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4812430 c.859G>C p.E287Q Chr.16: 28482379-28482379 on Build GRCh38
1301803 c.910G>C p.E304Q Chr.16: 28482379-28482379 on Build GRCh38
4513359 c.935C>T p.S312F Chr.16: 28482354-28482354 on Build GRCh38
4513360 c.884C>T p.S295F Chr.16: 28482354-28482354 on Build GRCh38
3508361 c.920T>G p.F307C Chr.16: 28482369-28482369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752882126 Chr.16: 28482249-28482249 on Build GRCh38
rs141816714 Chr.16: 28482351-28482351 on Build GRCh38
rs758705119 Chr.16: 28482306-28482306 on Build GRCh38
rs796052335 Chr.16: 28482340-28482340 on Build GRCh38
rs386833740 Chr.16: 28482344-28482345 on Build GRCh38
rs769358990 Chr.16: 28482348-28482348 on Build GRCh38
rs764357870 Chr.16: 28482385-28482385 on Build GRCh38
rs750876729 Chr.16: 28482301-28482301 on Build GRCh38
rs746276318 Chr.16: 28482213-28482213 on Build GRCh38
rs746616613 Chr.16: 28482215-28482215 on Build GRCh38
rs759304422 Chr.16: 28482374-28482374 on Build GRCh38
rs878945493 Chr.16: 28482258-28482258 on Build GRCh38
rs765319085 Chr.16: 28482225-28482225 on Build GRCh38
rs780974004 Chr.16: 28482343-28482343 on Build GRCh38
rs555927970 Chr.16: 28482275-28482275 on Build GRCh38
rs962144109 Chr.16: 28482258-28482258 on Build GRCh38
rs747723206 Chr.16: 28482344-28482344 on Build GRCh38
rs1015043618 Chr.16: 28482229-28482229 on Build GRCh38
rs368113837 Chr.16: 28482389-28482389 on Build GRCh38
rs762896453 Chr.16: 28482353-28482353 on Build GRCh38
rs757315798 Chr.16: 28482293-28482293 on Build GRCh38
rs770660659 Chr.16: 28482358-28482358 on Build GRCh38
rs1039413311 Chr.16: 28482388-28482388 on Build GRCh38
rs767096940 Chr.16: 28482273-28482273 on Build GRCh38
rs764268184 Chr.16: 28482290-28482290 on Build GRCh38
rs746674736 Chr.16: 28482325-28482325 on Build GRCh38
rs762657279 Chr.16: 28482242-28482242 on Build GRCh38
rs780419236 Chr.16: 28482315-28482315 on Build GRCh38
rs200023908 Chr.16: 28482214-28482214 on Build GRCh38
rs374019013 Chr.16: 28482294-28482294 on Build GRCh38
rs769259414 Chr.16: 28482213-28482213 on Build GRCh38
rs774239875 Chr.16: 28482373-28482373 on Build GRCh38
rs796052336 Chr.16: 28482324-28482324 on Build GRCh38
rs553687642 Chr.16: 28482223-28482223 on Build GRCh38
rs927361831 Chr.16: 28482266-28482266 on Build GRCh38
rs768466445 Chr.16: 28482331-28482331 on Build GRCh38
rs138636923 Chr.16: 28482330-28482330 on Build GRCh38
rs201457990 Chr.16: 28482291-28482291 on Build GRCh38
rs201945780 Chr.16: 28482233-28482233 on Build GRCh38
rs1057516267 Chr.16: 28482326-28482326 on Build GRCh38
rs766137609 Chr.16: 28482248-28482248 on Build GRCh38
rs542463265 Chr.16: 28482252-28482252 on Build GRCh38
rs773192286 Chr.16: 28482226-28482226 on Build GRCh38
rs545040014 Chr.16: 28482308-28482308 on Build GRCh38
rs971471454 Chr.16: 28482378-28482378 on Build GRCh38

View Full Product Details