Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33998432-33998691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMACR
Entrez Gene ID
23600
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
AMACRD, CBAS4, RACE, RM
UniGene
Hs.508343
Gene Chromosome Location
Chr.5: 33986986-34008115 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598987 c.559C>A p.L187I Chr.5: 33998660-33998660 on Build GRCh38
1283392 c.729G>T p.L243L Chr.5: 33998651-33998651 on Build GRCh38
6598986 c.720C>A p.F240L Chr.5: 33998660-33998660 on Build GRCh38
4381926 c.556G>T p.V186F Chr.5: 33998663-33998663 on Build GRCh38
5663483 c.542G>A p.S181N Chr.5: 33998677-33998677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753425865 Chr.5: 33998593-33998593 on Build GRCh38
rs199576063 Chr.5: 33998624-33998624 on Build GRCh38
rs769222146 Chr.5: 33998602-33998602 on Build GRCh38
rs200824585 Chr.5: 33998666-33998666 on Build GRCh38
rs1047555585 Chr.5: 33998509-33998509 on Build GRCh38
rs752535349 Chr.5: 33998648-33998648 on Build GRCh38
rs1005713559 Chr.5: 33998553-33998553 on Build GRCh38
rs767427040 Chr.5: 33998640-33998640 on Build GRCh38
rs375868211 Chr.5: 33998612-33998612 on Build GRCh38
rs376704656 Chr.5: 33998625-33998625 on Build GRCh38
rs961657958 Chr.5: 33998563-33998563 on Build GRCh38
rs778441339 Chr.5: 33998595-33998595 on Build GRCh38
rs890054237 Chr.5: 33998484-33998484 on Build GRCh38
rs537833252 Chr.5: 33998626-33998626 on Build GRCh38
rs1019652533 Chr.5: 33998548-33998548 on Build GRCh38
rs113345038 Chr.5: 33998659-33998659 on Build GRCh38
rs529626592 Chr.5: 33998589-33998589 on Build GRCh38
rs1013555111 Chr.5: 33998586-33998586 on Build GRCh38
rs763906079 Chr.5: 33998656-33998656 on Build GRCh38
rs34677 Chr.5: 33998663-33998663 on Build GRCh38
rs112753010 Chr.5: 33998630-33998630 on Build GRCh38
rs560448241 Chr.5: 33998580-33998580 on Build GRCh38
rs184782882 Chr.5: 33998603-33998603 on Build GRCh38
rs1039736668 Chr.5: 33998481-33998481 on Build GRCh38
rs201659164 Chr.5: 33998658-33998658 on Build GRCh38
rs551357353 Chr.5: 33998651-33998651 on Build GRCh38
rs774630103 Chr.5: 33998467-33998467 on Build GRCh38
rs748386474 Chr.5: 33998594-33998594 on Build GRCh38
rs778295213 Chr.5: 33998667-33998667 on Build GRCh38
rs747535789 Chr.5: 33998599-33998599 on Build GRCh38
rs9282594 Chr.5: 33998668-33998668 on Build GRCh38
rs369971392 Chr.5: 33998623-33998623 on Build GRCh38
rs1017038629 Chr.5: 33998571-33998571 on Build GRCh38
rs547192905 Chr.5: 33998505-33998505 on Build GRCh38
rs748512651 Chr.5: 33998610-33998610 on Build GRCh38

View Full Product Details