Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70835410-70835637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD207
Entrez Gene ID
50489
Gene Name
CD207 molecule
Gene Aliases
CLEC4K
UniGene
Hs.199731
Gene Chromosome Location
Chr.2: 70825248-70860787 on Build GRCh38
Transcripts
NM_015717.4 XM_011532874.1 XM_011532875.2 XM_011532876.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6542697 c.163G>A p.A55T Chr.2: 70835518-70835518 on Build GRCh38
6542698 c.150G>A p.T50T Chr.2: 70835531-70835531 on Build GRCh38
350666 c.119C>A p.P40H Chr.2: 70835562-70835562 on Build GRCh38
5862776 c.106C>T p.P36S Chr.2: 70835575-70835575 on Build GRCh38
148875 c.164C>T p.A55V Chr.2: 70835517-70835517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782385053 Chr.2: 70835579-70835579 on Build GRCh38
rs10489990 Chr.2: 70835517-70835517 on Build GRCh38
rs782316540 Chr.2: 70835509-70835509 on Build GRCh38
rs76379751 Chr.2: 70835470-70835470 on Build GRCh38
rs782546321 Chr.2: 70835456-70835456 on Build GRCh38
rs377312536 Chr.2: 70835563-70835563 on Build GRCh38
rs782566001 Chr.2: 70835561-70835561 on Build GRCh38
rs368757763 Chr.2: 70835562-70835562 on Build GRCh38
rs782229632 Chr.2: 70835439-70835439 on Build GRCh38
rs781823291 Chr.2: 70835546-70835546 on Build GRCh38
rs781788289 Chr.2: 70835468-70835468 on Build GRCh38
rs373401682 Chr.2: 70835599-70835599 on Build GRCh38
rs782200265 Chr.2: 70835564-70835564 on Build GRCh38
rs782078673 Chr.2: 70835597-70835597 on Build GRCh38
rs782013427 Chr.2: 70835581-70835581 on Build GRCh38
rs973709836 Chr.2: 70835440-70835440 on Build GRCh38
rs201227431 Chr.2: 70835498-70835498 on Build GRCh38
rs201606068 Chr.2: 70835513-70835513 on Build GRCh38
rs781983818 Chr.2: 70835518-70835518 on Build GRCh38
rs781863029 Chr.2: 70835453-70835453 on Build GRCh38
rs200240147 Chr.2: 70835497-70835497 on Build GRCh38
rs782673601 Chr.2: 70835464-70835464 on Build GRCh38
rs373616341 Chr.2: 70835539-70835539 on Build GRCh38
rs782778641 Chr.2: 70835532-70835532 on Build GRCh38
rs782508063 Chr.2: 70835550-70835550 on Build GRCh38
rs782342629 Chr.2: 70835569-70835569 on Build GRCh38
rs372245977 Chr.2: 70835590-70835590 on Build GRCh38
rs368255137 Chr.2: 70835573-70835573 on Build GRCh38
rs1017082055 Chr.2: 70835585-70835585 on Build GRCh38
rs370757828 Chr.2: 70835519-70835519 on Build GRCh38
rs112095621 Chr.2: 70835543-70835543 on Build GRCh38
rs76024692 Chr.2: 70835591-70835591 on Build GRCh38
rs782771176 Chr.2: 70835441-70835441 on Build GRCh38
rs555101184 Chr.2: 70835434-70835434 on Build GRCh38
rs782391335 Chr.2: 70835474-70835474 on Build GRCh38
rs372230506 Chr.2: 70835554-70835554 on Build GRCh38
rs79932602 Chr.2: 70835594-70835594 on Build GRCh38
rs782242067 Chr.2: 70835574-70835574 on Build GRCh38
rs374036641 Chr.2: 70835471-70835471 on Build GRCh38
rs562632122 Chr.2: 70835545-70835545 on Build GRCh38
rs781939546 Chr.2: 70835512-70835512 on Build GRCh38

View Full Product Details