Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90278389-90278616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRDE2
Entrez Gene ID
55051
Gene Name
NRDE-2, necessary for RNA interference, domain containing
Gene Aliases
C14orf102
UniGene
Hs.528131
Gene Chromosome Location
Chr.14: 90274276-90332137 on Build GRCh38
Transcripts
NM_017970.3 XM_011536886.1 XM_017021394.1 XM_017021395.1 XM_017021396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7380313 c.3385G>C p.A1129P Chr.14: 90278446-90278446 on Build GRCh38
7281658 c.3370-4delC p.? Chr.14: 90278465-90278465 on Build GRCh38
4488930 c.3400C>T p.P1134S Chr.14: 90278431-90278431 on Build GRCh38
5766548 c.3420C>T p.I1140I Chr.14: 90278411-90278411 on Build GRCh38
6729511 c.3387C>T p.A1129A Chr.14: 90278444-90278444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050406151 Chr.14: 90278518-90278518 on Build GRCh38
rs114392198 Chr.14: 90278496-90278496 on Build GRCh38
rs776805454 Chr.14: 90278412-90278412 on Build GRCh38
rs562566492 Chr.14: 90278486-90278486 on Build GRCh38
rs769831519 Chr.14: 90278469-90278469 on Build GRCh38
rs749144643 Chr.14: 90278538-90278538 on Build GRCh38
rs71853041 Chr.14: 90278515-90278517 on Build GRCh38
rs754934864 Chr.14: 90278497-90278497 on Build GRCh38
rs368982749 Chr.14: 90278547-90278547 on Build GRCh38
rs1051125711 Chr.14: 90278414-90278414 on Build GRCh38
rs201104859 Chr.14: 90278420-90278420 on Build GRCh38
rs201062537 Chr.14: 90278432-90278432 on Build GRCh38
rs771196098 Chr.14: 90278432-90278433 on Build GRCh38
rs765508701 Chr.14: 90278418-90278418 on Build GRCh38
rs191562667 Chr.14: 90278508-90278508 on Build GRCh38
rs993951466 Chr.14: 90278576-90278576 on Build GRCh38
rs917507955 Chr.14: 90278488-90278488 on Build GRCh38
rs901091901 Chr.14: 90278575-90278575 on Build GRCh38
rs370954669 Chr.14: 90278444-90278444 on Build GRCh38
rs753403812 Chr.14: 90278438-90278438 on Build GRCh38
rs750217500 Chr.14: 90278429-90278429 on Build GRCh38
rs148914817 Chr.14: 90278470-90278470 on Build GRCh38
rs764377000 Chr.14: 90278481-90278481 on Build GRCh38
rs574617605 Chr.14: 90278567-90278567 on Build GRCh38
rs934702649 Chr.14: 90278585-90278585 on Build GRCh38
rs760491210 Chr.14: 90278484-90278484 on Build GRCh38
rs372625671 Chr.14: 90278519-90278521 on Build GRCh38
rs747048338 Chr.14: 90278423-90278423 on Build GRCh38
rs377201570 Chr.14: 90278428-90278428 on Build GRCh38
rs771452952 Chr.14: 90278447-90278447 on Build GRCh38
rs745963878 Chr.14: 90278467-90278467 on Build GRCh38
rs763231863 Chr.14: 90278425-90278425 on Build GRCh38
rs921162644 Chr.14: 90278536-90278536 on Build GRCh38
rs139694903 Chr.14: 90278443-90278443 on Build GRCh38
rs763311441 Chr.14: 90278476-90278476 on Build GRCh38
rs776488985 Chr.14: 90278446-90278446 on Build GRCh38
rs775507521 Chr.14: 90278422-90278422 on Build GRCh38
rs1009334031 Chr.14: 90278553-90278553 on Build GRCh38
rs774693085 Chr.14: 90278464-90278465 on Build GRCh38
rs542643460 Chr.14: 90278483-90278483 on Build GRCh38
rs771398056 Chr.14: 90278410-90278410 on Build GRCh38
rs924675306 Chr.14: 90278570-90278570 on Build GRCh38
rs776943759 Chr.14: 90278452-90278452 on Build GRCh38

View Full Product Details