Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15845914-15846145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S2
Entrez Gene ID
8905
Gene Name
adaptor related protein complex 1 sigma 2 subunit
Gene Aliases
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B
UniGene
Hs.121592
Gene Chromosome Location
Chr.X: 15825806-15855014 on Build GRCh38
Transcripts
NM_001272071.1 NM_003916.4 XM_005274614.3 XM_011545599.1 XM_017029925.1 XM_017029926.1

SNP Detail

SNP ID Chromosome Location
rs912444495 Chr.X: 15845951-15845951 on Build GRCh38
rs916811795 Chr.X: 15846106-15846106 on Build GRCh38
rs137852213 Chr.X: 15845965-15845965 on Build GRCh38
rs755082763 Chr.X: 15845963-15845963 on Build GRCh38
rs773621681 Chr.X: 15846057-15846057 on Build GRCh38
rs989710564 Chr.X: 15846088-15846088 on Build GRCh38
rs587776738 Chr.X: 15846013-15846016 on Build GRCh38
rs192172204 Chr.X: 15846016-15846016 on Build GRCh38
rs942621863 Chr.X: 15845954-15845954 on Build GRCh38
rs747043868 Chr.X: 15845943-15845943 on Build GRCh38
rs763717703 Chr.X: 15846094-15846095 on Build GRCh38
rs781486489 Chr.X: 15846008-15846008 on Build GRCh38
rs781027153 Chr.X: 15846033-15846033 on Build GRCh38
rs770262016 Chr.X: 15846024-15846024 on Build GRCh38

View Full Product Details