Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50480704-50480969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC10
Entrez Gene ID
284361
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
C19orf63, HSM1, HSS1
UniGene
Hs.448941
Gene Chromosome Location
Chr.19: 50476477-50505905 on Build GRCh38
Transcripts
NM_175063.5 NM_206538.3 XM_006723163.3 XM_011526818.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4878701 c.566C>T p.P189L Chr.19: 50480744-50480744 on Build GRCh38
1395500 c.610C>T p.R204C Chr.19: 50480909-50480909 on Build GRCh38
4080409 c.538C>T p.L180L Chr.19: 50480716-50480716 on Build GRCh38
3713021 c.583G>T p.G195C Chr.19: 50480761-50480761 on Build GRCh38
440052 c.551C>T p.S184L Chr.19: 50480729-50480729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199665445 Chr.19: 50480918-50480918 on Build GRCh38
rs781541023 Chr.19: 50480836-50480836 on Build GRCh38
rs767029184 Chr.19: 50480886-50480886 on Build GRCh38
rs372648694 Chr.19: 50480731-50480731 on Build GRCh38
rs373717670 Chr.19: 50480895-50480895 on Build GRCh38
rs776598448 Chr.19: 50480861-50480863 on Build GRCh38
rs10410120 Chr.19: 50480745-50480745 on Build GRCh38
rs964920780 Chr.19: 50480887-50480887 on Build GRCh38
rs765460348 Chr.19: 50480796-50480796 on Build GRCh38
rs780301877 Chr.19: 50480804-50480804 on Build GRCh38
rs778889874 Chr.19: 50480927-50480927 on Build GRCh38
rs543448718 Chr.19: 50480805-50480805 on Build GRCh38
rs770834173 Chr.19: 50480847-50480848 on Build GRCh38
rs201394073 Chr.19: 50480892-50480892 on Build GRCh38
rs757540220 Chr.19: 50480733-50480733 on Build GRCh38
rs773262366 Chr.19: 50480781-50480781 on Build GRCh38
rs772989238 Chr.19: 50480726-50480726 on Build GRCh38
rs148959167 Chr.19: 50480739-50480739 on Build GRCh38
rs143536282 Chr.19: 50480744-50480744 on Build GRCh38
rs771613772 Chr.19: 50480944-50480944 on Build GRCh38
rs775272055 Chr.19: 50480869-50480869 on Build GRCh38
rs754977084 Chr.19: 50480812-50480812 on Build GRCh38
rs777480440 Chr.19: 50480845-50480845 on Build GRCh38
rs754182565 Chr.19: 50480732-50480732 on Build GRCh38
rs754830010 Chr.19: 50480817-50480817 on Build GRCh38
rs374258787 Chr.19: 50480948-50480948 on Build GRCh38
rs982050200 Chr.19: 50480785-50480785 on Build GRCh38
rs768950243 Chr.19: 50480755-50480755 on Build GRCh38
rs769468914 Chr.19: 50480777-50480777 on Build GRCh38
rs369893864 Chr.19: 50480729-50480729 on Build GRCh38
rs200217945 Chr.19: 50480909-50480909 on Build GRCh38
rs773663513 Chr.19: 50480884-50480884 on Build GRCh38
rs548942090 Chr.19: 50480787-50480787 on Build GRCh38
rs750963039 Chr.19: 50480742-50480742 on Build GRCh38
rs748010782 Chr.19: 50480762-50480762 on Build GRCh38
rs745422496 Chr.19: 50480858-50480858 on Build GRCh38
rs746471475 Chr.19: 50480812-50480812 on Build GRCh38
rs770437098 Chr.19: 50480783-50480783 on Build GRCh38
rs759126769 Chr.19: 50480730-50480730 on Build GRCh38
rs762307083 Chr.19: 50480788-50480788 on Build GRCh38
rs897381143 Chr.19: 50480878-50480878 on Build GRCh38
rs546645723 Chr.19: 50480829-50480829 on Build GRCh38
rs371056248 Chr.19: 50480852-50480852 on Build GRCh38
rs765664334 Chr.19: 50480883-50480883 on Build GRCh38
rs370619591 Chr.19: 50480893-50480893 on Build GRCh38
rs143690196 Chr.19: 50480896-50480896 on Build GRCh38
rs745782005 Chr.19: 50480936-50480936 on Build GRCh38
rs748301894 Chr.19: 50480837-50480837 on Build GRCh38
rs751735018 Chr.19: 50480809-50480809 on Build GRCh38
rs749131925 Chr.19: 50480910-50480910 on Build GRCh38
rs539247639 Chr.19: 50480876-50480876 on Build GRCh38
rs374288851 Chr.19: 50480866-50480866 on Build GRCh38
rs375662086 Chr.19: 50480786-50480786 on Build GRCh38
rs374611840 Chr.19: 50480847-50480847 on Build GRCh38
rs528586712 Chr.19: 50480819-50480819 on Build GRCh38
rs890150756 Chr.19: 50480846-50480846 on Build GRCh38
rs773373720 Chr.19: 50480751-50480751 on Build GRCh38
rs759210461 Chr.19: 50480784-50480784 on Build GRCh38
rs762514069 Chr.19: 50480727-50480727 on Build GRCh38
rs201206109 Chr.19: 50480848-50480848 on Build GRCh38
rs756253680 Chr.19: 50480901-50480901 on Build GRCh38
rs762821862 Chr.19: 50480782-50480782 on Build GRCh38
rs888541956 Chr.19: 50480905-50480905 on Build GRCh38
rs781245066 Chr.19: 50480761-50480761 on Build GRCh38
rs763387050 Chr.19: 50480885-50480885 on Build GRCh38
rs750967424 Chr.19: 50480798-50480798 on Build GRCh38
rs755954007 Chr.19: 50480839-50480839 on Build GRCh38
rs770809510 Chr.19: 50480851-50480851 on Build GRCh38
rs202184639 Chr.19: 50480803-50480803 on Build GRCh38

View Full Product Details