Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161123736-161123973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEDD
Entrez Gene ID
9191
Gene Name
death effector domain containing
Gene Aliases
CASP8IP1, DEDD1, DEFT, FLDED1, KE05
UniGene
Hs.744092
Gene Chromosome Location
Chr.1: 161120977-161132783 on Build GRCh38
Transcripts
NM_001039711.1 NM_001039712.1 NM_032998.2 XM_005245597.3 XM_005245598.4 XM_005245599.2 XM_005245600.2 XM_017002802.1 NM_001185092.1 XM_006711341.3 XM_017001371.1

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
-
UniGene
Hs.146406
Gene Chromosome Location
Chr.1: 161118072-161125446 on Build GRCh38
Transcripts
NM_001039711.1 NM_001039712.1 NM_032998.2 XM_005245597.3 XM_005245598.4 XM_005245599.2 XM_005245600.2 XM_017002802.1 NM_001185092.1 XM_006711341.3 XM_017001371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5041302 c.341T>C p.V114A Chr.1: 161123931-161123931 on Build GRCh38
4544386 c.355G>A p.E119K Chr.1: 161123917-161123917 on Build GRCh38
6333354 c.407C>A p.P136Q Chr.1: 161123865-161123865 on Build GRCh38
898471 c.372C>T p.R124R Chr.1: 161123900-161123900 on Build GRCh38
463270 c.354G>T p.L118L Chr.1: 161123918-161123918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753218307 Chr.1: 161123906-161123906 on Build GRCh38
rs755487788 Chr.1: 161123789-161123789 on Build GRCh38
rs749533503 Chr.1: 161123821-161123821 on Build GRCh38
rs933198992 Chr.1: 161123881-161123881 on Build GRCh38
rs775597637 Chr.1: 161123860-161123860 on Build GRCh38
rs761160550 Chr.1: 161123915-161123915 on Build GRCh38
rs909080200 Chr.1: 161123853-161123853 on Build GRCh38
rs769466281 Chr.1: 161123896-161123896 on Build GRCh38
rs572205717 Chr.1: 161123899-161123899 on Build GRCh38
rs759977685 Chr.1: 161123902-161123902 on Build GRCh38
rs75250474 Chr.1: 161123861-161123861 on Build GRCh38
rs764229115 Chr.1: 161123924-161123924 on Build GRCh38
rs917546503 Chr.1: 161123777-161123777 on Build GRCh38
rs768034686 Chr.1: 161123792-161123792 on Build GRCh38
rs778618227 Chr.1: 161123849-161123849 on Build GRCh38
rs757465896 Chr.1: 161123932-161123932 on Build GRCh38
rs771941069 Chr.1: 161123859-161123859 on Build GRCh38
rs754397500 Chr.1: 161123802-161123804 on Build GRCh38
rs772691525 Chr.1: 161123897-161123897 on Build GRCh38
rs768292725 Chr.1: 161123872-161123872 on Build GRCh38
rs372926625 Chr.1: 161123925-161123925 on Build GRCh38
rs1010357582 Chr.1: 161123900-161123900 on Build GRCh38
rs767805103 Chr.1: 161123905-161123905 on Build GRCh38
rs753163198 Chr.1: 161123817-161123817 on Build GRCh38
rs776189817 Chr.1: 161123879-161123879 on Build GRCh38
rs371823914 Chr.1: 161123820-161123820 on Build GRCh38
rs757501168 Chr.1: 161123830-161123830 on Build GRCh38
rs751948650 Chr.1: 161123788-161123788 on Build GRCh38
rs374736648 Chr.1: 161123892-161123892 on Build GRCh38
rs745649711 Chr.1: 161123855-161123855 on Build GRCh38

View Full Product Details