Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184166083-184166334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL3
Entrez Gene ID
1857
Gene Name
dishevelled segment polarity protein 3
Gene Aliases
DRS3
UniGene
Hs.388116
Gene Chromosome Location
Chr.3: 184155311-184173614 on Build GRCh38
Transcripts
NM_004423.3 XM_005247172.2 XM_011512513.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6346790 c.780G>T p.L260F Chr.3: 184166142-184166142 on Build GRCh38
337848 c.849G>T p.K283N Chr.3: 184166211-184166211 on Build GRCh38
6836231 c.863C>A p.A288D Chr.3: 184166225-184166225 on Build GRCh38
5418350 c.851G>C p.G284A Chr.3: 184166213-184166213 on Build GRCh38
5743010 c.874C>T p.R292C Chr.3: 184166236-184166236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747297916 Chr.3: 184166237-184166237 on Build GRCh38
rs770286093 Chr.3: 184166277-184166277 on Build GRCh38
rs760740734 Chr.3: 184166299-184166316 on Build GRCh38
rs371458501 Chr.3: 184166207-184166207 on Build GRCh38
rs772607453 Chr.3: 184166174-184166174 on Build GRCh38
rs377251528 Chr.3: 184166286-184166286 on Build GRCh38
rs746045622 Chr.3: 184166118-184166118 on Build GRCh38
rs368504471 Chr.3: 184166311-184166311 on Build GRCh38
rs780121041 Chr.3: 184166234-184166234 on Build GRCh38
rs537851526 Chr.3: 184166145-184166145 on Build GRCh38
rs367769683 Chr.3: 184166181-184166181 on Build GRCh38
rs755336198 Chr.3: 184166242-184166242 on Build GRCh38
rs200438406 Chr.3: 184166309-184166309 on Build GRCh38
rs748593506 Chr.3: 184166275-184166275 on Build GRCh38
rs559178975 Chr.3: 184166291-184166291 on Build GRCh38
rs764433293 Chr.3: 184166176-184166176 on Build GRCh38
rs879163426 Chr.3: 184166218-184166218 on Build GRCh38
rs142819846 Chr.3: 184166220-184166220 on Build GRCh38
rs142071816 Chr.3: 184166173-184166173 on Build GRCh38
rs199853617 Chr.3: 184166182-184166182 on Build GRCh38
rs746880708 Chr.3: 184166269-184166269 on Build GRCh38
rs941537503 Chr.3: 184166199-184166199 on Build GRCh38
rs780427517 Chr.3: 184166122-184166122 on Build GRCh38
rs762913408 Chr.3: 184166313-184166313 on Build GRCh38
rs987348133 Chr.3: 184166126-184166126 on Build GRCh38
rs748767904 Chr.3: 184166280-184166280 on Build GRCh38
rs11925802 Chr.3: 184166284-184166284 on Build GRCh38
rs896517683 Chr.3: 184166270-184166270 on Build GRCh38
rs201788751 Chr.3: 184166214-184166214 on Build GRCh38
rs781598570 Chr.3: 184166271-184166271 on Build GRCh38
rs549144110 Chr.3: 184166102-184166102 on Build GRCh38
rs769700978 Chr.3: 184166114-184166114 on Build GRCh38
rs138011240 Chr.3: 184166169-184166169 on Build GRCh38
rs375463143 Chr.3: 184166304-184166304 on Build GRCh38
rs757381531 Chr.3: 184166106-184166106 on Build GRCh38
rs373868630 Chr.3: 184166120-184166120 on Build GRCh38
rs765975448 Chr.3: 184166178-184166178 on Build GRCh38
rs759327886 Chr.3: 184166287-184166287 on Build GRCh38
rs758598662 Chr.3: 184166232-184166232 on Build GRCh38
rs747357473 Chr.3: 184166123-184166123 on Build GRCh38
rs777094418 Chr.3: 184166151-184166151 on Build GRCh38
rs539349424 Chr.3: 184166302-184166302 on Build GRCh38
rs779200044 Chr.3: 184166108-184166108 on Build GRCh38
rs750502107 Chr.3: 184166221-184166221 on Build GRCh38
rs372122546 Chr.3: 184166278-184166278 on Build GRCh38
rs201275800 Chr.3: 184166239-184166239 on Build GRCh38
rs767460283 Chr.3: 184166196-184166196 on Build GRCh38
rs918909756 Chr.3: 184166104-184166104 on Build GRCh38
rs1056470101 Chr.3: 184166241-184166241 on Build GRCh38

View Full Product Details