Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157575490-157575763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL4
Entrez Gene ID
83417
Gene Name
Fc receptor like 4
Gene Aliases
CD307d, FCRH4, IGFP2, IRTA1
UniGene
Hs.120260
Gene Chromosome Location
Chr.1: 157573749-157598080 on Build GRCh38
Transcripts
NM_031282.2 XM_011510034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
897543 c.1540G>T p.E514* Chr.1: 157575532-157575532 on Build GRCh38
4023510 c.1543A>G p.S515G Chr.1: 157575529-157575529 on Build GRCh38
5531180 c.1537G>A p.E513K Chr.1: 157575535-157575535 on Build GRCh38
5036008 c.1508A>C p.N503T Chr.1: 157575564-157575564 on Build GRCh38
1985179 c.1526G>T p.S509I Chr.1: 157575546-157575546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779927177 Chr.1: 157575664-157575664 on Build GRCh38
rs374476793 Chr.1: 157575731-157575731 on Build GRCh38
rs769920992 Chr.1: 157575549-157575549 on Build GRCh38
rs370866195 Chr.1: 157575562-157575562 on Build GRCh38
rs3811028 Chr.1: 157575594-157575594 on Build GRCh38
rs760191054 Chr.1: 157575630-157575630 on Build GRCh38
rs1026860925 Chr.1: 157575654-157575654 on Build GRCh38
rs201762910 Chr.1: 157575544-157575544 on Build GRCh38
rs887548684 Chr.1: 157575659-157575659 on Build GRCh38
rs767832134 Chr.1: 157575599-157575599 on Build GRCh38
rs762228769 Chr.1: 157575705-157575705 on Build GRCh38
rs367654161 Chr.1: 157575737-157575737 on Build GRCh38
rs1007989153 Chr.1: 157575578-157575578 on Build GRCh38
rs151244603 Chr.1: 157575565-157575565 on Build GRCh38
rs748415070 Chr.1: 157575668-157575668 on Build GRCh38
rs774804922 Chr.1: 157575566-157575566 on Build GRCh38
rs140634688 Chr.1: 157575693-157575693 on Build GRCh38
rs187311182 Chr.1: 157575521-157575521 on Build GRCh38
rs376151318 Chr.1: 157575627-157575627 on Build GRCh38
rs759973261 Chr.1: 157575701-157575701 on Build GRCh38
rs758299870 Chr.1: 157575655-157575655 on Build GRCh38
rs867211538 Chr.1: 157575517-157575517 on Build GRCh38
rs764388028 Chr.1: 157575537-157575537 on Build GRCh38
rs368411125 Chr.1: 157575637-157575637 on Build GRCh38
rs200200324 Chr.1: 157575574-157575574 on Build GRCh38
rs767525096 Chr.1: 157575710-157575710 on Build GRCh38
rs750338031 Chr.1: 157575643-157575643 on Build GRCh38
rs772443837 Chr.1: 157575707-157575707 on Build GRCh38
rs778048886 Chr.1: 157575679-157575679 on Build GRCh38
rs749363901 Chr.1: 157575689-157575689 on Build GRCh38
rs745699296 Chr.1: 157575527-157575527 on Build GRCh38
rs763790928 Chr.1: 157575631-157575631 on Build GRCh38
rs769965358 Chr.1: 157575671-157575671 on Build GRCh38
rs779042776 Chr.1: 157575641-157575641 on Build GRCh38
rs756761107 Chr.1: 157575638-157575638 on Build GRCh38
rs778991509 Chr.1: 157575526-157575526 on Build GRCh38
rs781587266 Chr.1: 157575536-157575536 on Build GRCh38
rs774857897 Chr.1: 157575699-157575699 on Build GRCh38
rs769507733 Chr.1: 157575538-157575538 on Build GRCh38
rs771971609 Chr.1: 157575528-157575528 on Build GRCh38
rs748339295 Chr.1: 157575542-157575542 on Build GRCh38

View Full Product Details