Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40737872-40738109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
UniGene
Hs.511067
Gene Chromosome Location
Chr.15: 40735884-40755336 on Build GRCh38
Transcripts
NM_001304802.1 NM_001323894.1 NM_001323895.1 NM_001323896.1 NM_001323897.1 NM_018145.2 XM_011521755.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5383226 c.1125G>A p.Q375Q Chr.15: 40737965-40737965 on Build GRCh38
961292 c.1098C>G p.H366Q Chr.15: 40737992-40737992 on Build GRCh38
6476338 c.1097A>T p.H366L Chr.15: 40737993-40737993 on Build GRCh38
6767031 c.1075C>T p.Q359* Chr.15: 40738015-40738015 on Build GRCh38
6015699 c.1103T>G p.L368R Chr.15: 40737987-40737987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757993967 Chr.15: 40737923-40737923 on Build GRCh38
rs368610327 Chr.15: 40738059-40738059 on Build GRCh38
rs765296676 Chr.15: 40738068-40738069 on Build GRCh38
rs748146910 Chr.15: 40737947-40737947 on Build GRCh38
rs370226833 Chr.15: 40737956-40737956 on Build GRCh38
rs147774712 Chr.15: 40737913-40737913 on Build GRCh38
rs536849601 Chr.15: 40737912-40737912 on Build GRCh38
rs756123686 Chr.15: 40738042-40738042 on Build GRCh38
rs751265623 Chr.15: 40738025-40738025 on Build GRCh38
rs764521318 Chr.15: 40738002-40738002 on Build GRCh38
rs577185516 Chr.15: 40738071-40738071 on Build GRCh38
rs767621061 Chr.15: 40737974-40737974 on Build GRCh38
rs754655723 Chr.15: 40738026-40738026 on Build GRCh38
rs761812108 Chr.15: 40737949-40737949 on Build GRCh38
rs747658608 Chr.15: 40738035-40738035 on Build GRCh38
rs754170561 Chr.15: 40738003-40738003 on Build GRCh38
rs1046388789 Chr.15: 40737959-40737959 on Build GRCh38
rs141170560 Chr.15: 40737971-40737971 on Build GRCh38
rs376231723 Chr.15: 40738000-40738000 on Build GRCh38
rs148669324 Chr.15: 40738022-40738022 on Build GRCh38
rs771071312 Chr.15: 40737969-40737969 on Build GRCh38
rs77019569 Chr.15: 40737898-40737898 on Build GRCh38
rs376771038 Chr.15: 40738047-40738047 on Build GRCh38
rs754498140 Chr.15: 40737941-40737941 on Build GRCh38
rs778536855 Chr.15: 40738063-40738063 on Build GRCh38
rs772944423 Chr.15: 40737955-40737955 on Build GRCh38
rs775612458 Chr.15: 40738082-40738082 on Build GRCh38
rs780818826 Chr.15: 40737945-40737945 on Build GRCh38
rs916597630 Chr.15: 40738086-40738086 on Build GRCh38
rs772266820 Chr.15: 40738074-40738074 on Build GRCh38
rs11558808 Chr.15: 40738004-40738004 on Build GRCh38
rs757596064 Chr.15: 40738020-40738020 on Build GRCh38
rs182138409 Chr.15: 40737938-40737938 on Build GRCh38
rs780908773 Chr.15: 40738028-40738028 on Build GRCh38
rs759639779 Chr.15: 40737972-40737972 on Build GRCh38
rs928943502 Chr.15: 40737977-40737977 on Build GRCh38
rs751631114 Chr.15: 40738090-40738091 on Build GRCh38
rs779702744 Chr.15: 40737931-40737931 on Build GRCh38
rs1008603063 Chr.15: 40737902-40737902 on Build GRCh38
rs769842993 Chr.15: 40737950-40737950 on Build GRCh38

View Full Product Details