Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69153978-69154239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
UniGene
Hs.25377
Gene Chromosome Location
Chr.17: 69148007-69244815 on Build GRCh38
Transcripts
NM_080282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298869 c.3803G>A p.S1268N Chr.17: 69153993-69153993 on Build GRCh38
707464 c.3786G>T p.V1262V Chr.17: 69154235-69154235 on Build GRCh38
7218446 c.3787-1G>A p.? Chr.17: 69154010-69154010 on Build GRCh38
390527 c.3787-1G>C p.? Chr.17: 69154010-69154010 on Build GRCh38
983564 c.3794T>C p.L1265S Chr.17: 69154002-69154002 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1057220471 Chr.17: 69154102-69154102 on Build GRCh38
rs937019879 Chr.17: 69154141-69154144 on Build GRCh38
rs369400674 Chr.17: 69154173-69154173 on Build GRCh38
rs185372264 Chr.17: 69154115-69154115 on Build GRCh38
rs771427156 Chr.17: 69154011-69154011 on Build GRCh38
rs557508194 Chr.17: 69154170-69154170 on Build GRCh38
rs766333221 Chr.17: 69154195-69154195 on Build GRCh38
rs904434814 Chr.17: 69154142-69154142 on Build GRCh38
rs765488334 Chr.17: 69154045-69154045 on Build GRCh38
rs940079536 Chr.17: 69154079-69154079 on Build GRCh38
rs138422677 Chr.17: 69154206-69154206 on Build GRCh38
rs752674370 Chr.17: 69154199-69154199 on Build GRCh38
rs141590994 Chr.17: 69154010-69154010 on Build GRCh38
rs750541228 Chr.17: 69154050-69154050 on Build GRCh38
rs763104475 Chr.17: 69154059-69154059 on Build GRCh38
rs951613427 Chr.17: 69154123-69154123 on Build GRCh38
rs766837501 Chr.17: 69154104-69154104 on Build GRCh38
rs142909799 Chr.17: 69154209-69154209 on Build GRCh38
rs1007287662 Chr.17: 69154068-69154068 on Build GRCh38
rs555577333 Chr.17: 69154043-69154043 on Build GRCh38
rs376929280 Chr.17: 69154166-69154166 on Build GRCh38
rs577453947 Chr.17: 69154121-69154121 on Build GRCh38
rs769183262 Chr.17: 69154034-69154034 on Build GRCh38
rs928298431 Chr.17: 69154138-69154138 on Build GRCh38
rs750932909 Chr.17: 69154058-69154058 on Build GRCh38
rs1000081898 Chr.17: 69154151-69154151 on Build GRCh38
rs866432129 Chr.17: 69154174-69154174 on Build GRCh38
rs370574737 Chr.17: 69154183-69154183 on Build GRCh38
rs2886232 Chr.17: 69154035-69154035 on Build GRCh38
rs558944942 Chr.17: 69154114-69154114 on Build GRCh38
rs778354658 Chr.17: 69154003-69154003 on Build GRCh38
rs145891525 Chr.17: 69154130-69154130 on Build GRCh38
rs574002350 Chr.17: 69154083-69154083 on Build GRCh38
rs887131161 Chr.17: 69154062-69154062 on Build GRCh38
rs1048415730 Chr.17: 69154040-69154040 on Build GRCh38
rs770558049 Chr.17: 69154001-69154001 on Build GRCh38
rs750661377 Chr.17: 69154020-69154020 on Build GRCh38
rs776085553 Chr.17: 69154018-69154018 on Build GRCh38

View Full Product Details