Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127788521-127788760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK9
Entrez Gene ID
1025
Gene Name
cyclin dependent kinase 9
Gene Aliases
C-2k, CDC2L4, CTK1, PITALRE, TAK
UniGene
Hs.150423
Gene Chromosome Location
Chr.9: 127786026-127790782 on Build GRCh38
Transcripts
NM_001261.3 XM_017014184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625608 c.735C>T p.C245C Chr.9: 127788674-127788674 on Build GRCh38
4999039 c.1046A>G p.N349S Chr.9: 127788634-127788634 on Build GRCh38
5063626 c.619delG p.G207fs*69 Chr.9: 127788558-127788558 on Build GRCh38
1725286 c.663G>T p.E221D Chr.9: 127788602-127788602 on Build GRCh38
4926984 c.1076G>T p.S359I Chr.9: 127788664-127788664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758197793 Chr.9: 127788705-127788729 on Build GRCh38
rs932742068 Chr.9: 127788702-127788702 on Build GRCh38
rs11554104 Chr.9: 127788654-127788654 on Build GRCh38
rs775163113 Chr.9: 127788556-127788556 on Build GRCh38
rs368389511 Chr.9: 127788704-127788704 on Build GRCh38
rs779479066 Chr.9: 127788641-127788641 on Build GRCh38
rs749487652 Chr.9: 127788705-127788705 on Build GRCh38
rs765527912 Chr.9: 127788708-127788708 on Build GRCh38
rs3217745 Chr.9: 127788738-127788738 on Build GRCh38
rs991873132 Chr.9: 127788587-127788587 on Build GRCh38
rs770752687 Chr.9: 127788739-127788739 on Build GRCh38
rs747985794 Chr.9: 127788628-127788629 on Build GRCh38
rs141577691 Chr.9: 127788637-127788637 on Build GRCh38
rs763885651 Chr.9: 127788558-127788558 on Build GRCh38
rs576155177 Chr.9: 127788731-127788731 on Build GRCh38
rs566601726 Chr.9: 127788714-127788714 on Build GRCh38
rs750537619 Chr.9: 127788613-127788613 on Build GRCh38
rs759014029 Chr.9: 127788650-127788650 on Build GRCh38
rs137949299 Chr.9: 127788656-127788656 on Build GRCh38
rs774177908 Chr.9: 127788684-127788684 on Build GRCh38
rs200268442 Chr.9: 127788696-127788696 on Build GRCh38
rs551379074 Chr.9: 127788700-127788700 on Build GRCh38
rs775012342 Chr.9: 127788663-127788663 on Build GRCh38
rs760683797 Chr.9: 127788698-127788698 on Build GRCh38
rs779676185 Chr.9: 127788727-127788727 on Build GRCh38
rs755440676 Chr.9: 127788559-127788560 on Build GRCh38
rs753143249 Chr.9: 127788709-127788709 on Build GRCh38
rs767418586 Chr.9: 127788612-127788612 on Build GRCh38
rs761561096 Chr.9: 127788687-127788687 on Build GRCh38
rs891203107 Chr.9: 127788720-127788720 on Build GRCh38
rs140058017 Chr.9: 127788540-127788540 on Build GRCh38
rs374586573 Chr.9: 127788736-127788736 on Build GRCh38
rs766414012 Chr.9: 127788703-127788703 on Build GRCh38
rs778384628 Chr.9: 127788653-127788653 on Build GRCh38
rs373394121 Chr.9: 127788549-127788549 on Build GRCh38
rs773135587 Chr.9: 127788737-127788737 on Build GRCh38
rs778441641 Chr.9: 127788713-127788713 on Build GRCh38
rs10460 Chr.9: 127788632-127788632 on Build GRCh38
rs377243893 Chr.9: 127788674-127788674 on Build GRCh38
rs762673744 Chr.9: 127788557-127788557 on Build GRCh38
rs143718708 Chr.9: 127788638-127788638 on Build GRCh38
rs375012308 Chr.9: 127788697-127788697 on Build GRCh38
rs761794166 Chr.9: 127788590-127788590 on Build GRCh38
rs756177544 Chr.9: 127788617-127788617 on Build GRCh38
rs199764560 Chr.9: 127788728-127788728 on Build GRCh38
rs768177225 Chr.9: 127788675-127788675 on Build GRCh38
rs555741634 Chr.9: 127788560-127788560 on Build GRCh38
rs745340199 Chr.9: 127788546-127788546 on Build GRCh38
rs371033254 Chr.9: 127788564-127788564 on Build GRCh38
rs184616297 Chr.9: 127788635-127788635 on Build GRCh38
rs377539796 Chr.9: 127788584-127788584 on Build GRCh38
rs778663248 Chr.9: 127788560-127788560 on Build GRCh38
rs757703558 Chr.9: 127788719-127788719 on Build GRCh38

View Full Product Details