Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151458255-151458512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMT1
Entrez Gene ID
79624
Gene Name
acidic residue methyltransferase 1
Gene Aliases
C6orf211
UniGene
Hs.15929
Gene Chromosome Location
Chr.6: 151452258-151470101 on Build GRCh38
Transcripts
NM_001286562.1 NM_024573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616451 c.151G>A p.G51S Chr.6: 151458331-151458331 on Build GRCh38
1311672 c.163G>A p.E55K Chr.6: 151458343-151458343 on Build GRCh38
4473428 c.184C>T p.L62F Chr.6: 151458364-151458364 on Build GRCh38
5543764 c.283C>T p.Q95* Chr.6: 151458463-151458463 on Build GRCh38
6487525 c.293T>C p.L98P Chr.6: 151458473-151458473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923278317 Chr.6: 151458330-151458330 on Build GRCh38
rs200290360 Chr.6: 151458431-151458431 on Build GRCh38
rs759940406 Chr.6: 151458454-151458454 on Build GRCh38
rs754257555 Chr.6: 151458357-151458357 on Build GRCh38
rs981839094 Chr.6: 151458329-151458329 on Build GRCh38
rs142167376 Chr.6: 151458286-151458287 on Build GRCh38
rs760794434 Chr.6: 151458440-151458440 on Build GRCh38
rs779433586 Chr.6: 151458377-151458377 on Build GRCh38
rs753230490 Chr.6: 151458466-151458466 on Build GRCh38
rs577243722 Chr.6: 151458285-151458286 on Build GRCh38
rs745655939 Chr.6: 151458287-151458288 on Build GRCh38
rs769658405 Chr.6: 151458301-151458301 on Build GRCh38
rs766575308 Chr.6: 151458452-151458452 on Build GRCh38
rs774428182 Chr.6: 151458318-151458318 on Build GRCh38
rs759930192 Chr.6: 151458355-151458355 on Build GRCh38
rs369794464 Chr.6: 151458324-151458324 on Build GRCh38
rs900570044 Chr.6: 151458448-151458448 on Build GRCh38
rs560894715 Chr.6: 151458361-151458361 on Build GRCh38
rs749164072 Chr.6: 151458409-151458409 on Build GRCh38
rs17850732 Chr.6: 151458410-151458410 on Build GRCh38
rs756224604 Chr.6: 151458341-151458341 on Build GRCh38
rs748628466 Chr.6: 151458381-151458381 on Build GRCh38
rs368958688 Chr.6: 151458436-151458436 on Build GRCh38
rs764188645 Chr.6: 151458315-151458315 on Build GRCh38
rs200297357 Chr.6: 151458405-151458405 on Build GRCh38
rs767667204 Chr.6: 151458434-151458434 on Build GRCh38
rs775085657 Chr.6: 151458302-151458302 on Build GRCh38
rs983669449 Chr.6: 151458414-151458414 on Build GRCh38
rs769276679 Chr.6: 151458402-151458402 on Build GRCh38
rs140124563 Chr.6: 151458482-151458482 on Build GRCh38
rs147918835 Chr.6: 151458362-151458362 on Build GRCh38
rs563766286 Chr.6: 151458457-151458457 on Build GRCh38
rs758588286 Chr.6: 151458473-151458474 on Build GRCh38
rs35036943 Chr.6: 151458399-151458399 on Build GRCh38
rs959322933 Chr.6: 151458349-151458349 on Build GRCh38
rs755563017 Chr.6: 151458291-151458292 on Build GRCh38
rs745458126 Chr.6: 151458287-151458287 on Build GRCh38
rs779095524 Chr.6: 151458430-151458431 on Build GRCh38
rs1037911888 Chr.6: 151458400-151458400 on Build GRCh38
rs768541724 Chr.6: 151458415-151458415 on Build GRCh38
rs541327002 Chr.6: 151458342-151458342 on Build GRCh38
rs973084534 Chr.6: 151458376-151458376 on Build GRCh38
rs752828244 Chr.6: 151458467-151458468 on Build GRCh38
rs763001248 Chr.6: 151458308-151458308 on Build GRCh38
rs891754848 Chr.6: 151458467-151458467 on Build GRCh38
rs1054460758 Chr.6: 151458456-151458456 on Build GRCh38
rs756810057 Chr.6: 151458387-151458387 on Build GRCh38
rs767473583 Chr.6: 151458333-151458333 on Build GRCh38
rs774481276 Chr.6: 151458424-151458424 on Build GRCh38
rs754026626 Chr.6: 151458453-151458453 on Build GRCh38
rs750680617 Chr.6: 151458334-151458334 on Build GRCh38
rs750741996 Chr.6: 151458286-151458289 on Build GRCh38
rs747476047 Chr.6: 151458401-151458401 on Build GRCh38
rs765534550 Chr.6: 151458460-151458460 on Build GRCh38

View Full Product Details