Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138239257-138239520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
UniGene
Hs.744868
Gene Chromosome Location
Chr.3: 138187248-138298389 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_014154.4 NM_015396.5 NM_213654.3 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 XM_017006100.1 XM_017006101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5803014 c.782G>A p.C261Y Chr.3: 138239515-138239515 on Build GRCh38
5803015 c.782G>A p.C261Y Chr.3: 138239515-138239515 on Build GRCh38
318781 c.763A>T p.I255F Chr.3: 138239496-138239496 on Build GRCh38
5917967 c.735-3C>T p.? Chr.3: 138239465-138239465 on Build GRCh38
5917968 c.777-3C>T p.? Chr.3: 138239465-138239465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759750685 Chr.3: 138239454-138239457 on Build GRCh38
rs753249340 Chr.3: 138239480-138239480 on Build GRCh38
rs756865437 Chr.3: 138239490-138239490 on Build GRCh38
rs150444078 Chr.3: 138239310-138239310 on Build GRCh38
rs781779763 Chr.3: 138239477-138239477 on Build GRCh38
rs566510971 Chr.3: 138239328-138239328 on Build GRCh38
rs931869554 Chr.3: 138239347-138239347 on Build GRCh38
rs774443238 Chr.3: 138239437-138239438 on Build GRCh38
rs138278309 Chr.3: 138239340-138239340 on Build GRCh38
rs1036859359 Chr.3: 138239411-138239411 on Build GRCh38
rs1050280952 Chr.3: 138239377-138239377 on Build GRCh38
rs761949710 Chr.3: 138239422-138239422 on Build GRCh38
rs542059414 Chr.3: 138239459-138239459 on Build GRCh38
rs755558879 Chr.3: 138239473-138239473 on Build GRCh38
rs765201694 Chr.3: 138239423-138239423 on Build GRCh38
rs538958527 Chr.3: 138239432-138239432 on Build GRCh38
rs951925173 Chr.3: 138239346-138239346 on Build GRCh38
rs923673944 Chr.3: 138239322-138239322 on Build GRCh38
rs961428325 Chr.3: 138239338-138239338 on Build GRCh38
rs555474918 Chr.3: 138239360-138239360 on Build GRCh38
rs573360434 Chr.3: 138239341-138239341 on Build GRCh38
rs780009276 Chr.3: 138239482-138239482 on Build GRCh38
rs910355114 Chr.3: 138239441-138239441 on Build GRCh38
rs956581541 Chr.3: 138239336-138239336 on Build GRCh38
rs192191819 Chr.3: 138239415-138239415 on Build GRCh38
rs944534048 Chr.3: 138239387-138239387 on Build GRCh38
rs186906659 Chr.3: 138239293-138239293 on Build GRCh38
rs557383588 Chr.3: 138239378-138239378 on Build GRCh38
rs1025415165 Chr.3: 138239329-138239329 on Build GRCh38
rs751808182 Chr.3: 138239465-138239465 on Build GRCh38
rs201024459 Chr.3: 138239433-138239433 on Build GRCh38

View Full Product Details