Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102730353-102730610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC168
Entrez Gene ID
643677
Gene Name
coiled-coil domain containing 168
Gene Aliases
C13orf40
UniGene
Hs.368382
Gene Chromosome Location
Chr.13: 102729367-102759072 on Build GRCh38
Transcripts
NM_001146197.1 XM_011521106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554106 c.6290C>A p.P2097Q Chr.13: 102730520-102730520 on Build GRCh38
4475682 c.6205C>T p.L2069F Chr.13: 102730605-102730605 on Build GRCh38
5807447 c.6432A>T p.T2144T Chr.13: 102730378-102730378 on Build GRCh38
6538005 c.6282T>C p.H2094H Chr.13: 102730528-102730528 on Build GRCh38
5725740 c.6279C>T p.F2093F Chr.13: 102730531-102730531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920323636 Chr.13: 102730392-102730392 on Build GRCh38
rs774830202 Chr.13: 102730435-102730435 on Build GRCh38
rs745497329 Chr.13: 102730510-102730511 on Build GRCh38
rs757131569 Chr.13: 102730571-102730571 on Build GRCh38
rs1010318216 Chr.13: 102730574-102730574 on Build GRCh38
rs764166437 Chr.13: 102730499-102730499 on Build GRCh38
rs931946572 Chr.13: 102730530-102730530 on Build GRCh38
rs1042234043 Chr.13: 102730579-102730579 on Build GRCh38
rs893171730 Chr.13: 102730564-102730564 on Build GRCh38
rs193108690 Chr.13: 102730587-102730587 on Build GRCh38
rs775730782 Chr.13: 102730495-102730495 on Build GRCh38
rs532369433 Chr.13: 102730500-102730500 on Build GRCh38
rs117848272 Chr.13: 102730570-102730570 on Build GRCh38
rs373076604 Chr.13: 102730433-102730433 on Build GRCh38
rs771807941 Chr.13: 102730428-102730428 on Build GRCh38
rs771198253 Chr.13: 102730426-102730426 on Build GRCh38
rs763099124 Chr.13: 102730496-102730496 on Build GRCh38
rs866328930 Chr.13: 102730408-102730408 on Build GRCh38
rs565986583 Chr.13: 102730586-102730586 on Build GRCh38
rs376038536 Chr.13: 102730466-102730466 on Build GRCh38
rs776971686 Chr.13: 102730429-102730429 on Build GRCh38
rs1001326788 Chr.13: 102730559-102730559 on Build GRCh38
rs763888765 Chr.13: 102730479-102730479 on Build GRCh38
rs765456917 Chr.13: 102730451-102730451 on Build GRCh38
rs760261867 Chr.13: 102730453-102730453 on Build GRCh38
rs943944663 Chr.13: 102730409-102730409 on Build GRCh38
rs951783526 Chr.13: 102730395-102730395 on Build GRCh38
rs945346086 Chr.13: 102730471-102730471 on Build GRCh38
rs1054441559 Chr.13: 102730552-102730552 on Build GRCh38

View Full Product Details