Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31111279-31111512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDK
Entrez Gene ID
10295
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
BCKDKD, BDK
UniGene
Hs.513520
Gene Chromosome Location
Chr.16: 31108294-31117651 on Build GRCh38
Transcripts
NM_001122957.2 NM_001271926.1 NM_005881.3 XM_017022859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970122 c.922G>A p.D308N Chr.16: 31111376-31111376 on Build GRCh38
6610595 c.906C>T p.I302I Chr.16: 31111360-31111360 on Build GRCh38
6464343 c.853A>T p.M285L Chr.16: 31111307-31111307 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979865656 Chr.16: 31111417-31111417 on Build GRCh38
rs921395862 Chr.16: 31111318-31111318 on Build GRCh38
rs760851100 Chr.16: 31111301-31111301 on Build GRCh38
rs756951861 Chr.16: 31111356-31111356 on Build GRCh38
rs958246870 Chr.16: 31111312-31111312 on Build GRCh38
rs989757942 Chr.16: 31111333-31111333 on Build GRCh38
rs772364078 Chr.16: 31111426-31111426 on Build GRCh38
rs753857407 Chr.16: 31111321-31111321 on Build GRCh38
rs938450372 Chr.16: 31111486-31111486 on Build GRCh38
rs141506795 Chr.16: 31111418-31111418 on Build GRCh38
rs758133599 Chr.16: 31111366-31111366 on Build GRCh38
rs757272693 Chr.16: 31111360-31111360 on Build GRCh38
rs746453488 Chr.16: 31111376-31111376 on Build GRCh38
rs754517816 Chr.16: 31111398-31111398 on Build GRCh38
rs767540350 Chr.16: 31111401-31111401 on Build GRCh38
rs760797698 Chr.16: 31111430-31111430 on Build GRCh38
rs906230942 Chr.16: 31111472-31111472 on Build GRCh38
rs1002358085 Chr.16: 31111493-31111493 on Build GRCh38
rs774848215 Chr.16: 31111410-31111410 on Build GRCh38
rs577834584 Chr.16: 31111432-31111432 on Build GRCh38
rs200253972 Chr.16: 31111371-31111371 on Build GRCh38
rs771171406 Chr.16: 31111408-31111408 on Build GRCh38
rs765467214 Chr.16: 31111309-31111309 on Build GRCh38
rs371078966 Chr.16: 31111305-31111305 on Build GRCh38
rs765128280 Chr.16: 31111357-31111357 on Build GRCh38
rs1054014743 Chr.16: 31111361-31111361 on Build GRCh38
rs374408251 Chr.16: 31111399-31111399 on Build GRCh38
rs201164597 Chr.16: 31111358-31111358 on Build GRCh38

View Full Product Details