Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123383626-123383857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
UniGene
Hs.744995
Gene Chromosome Location
Chr.9: 123379654-123930158 on Build GRCh38
Transcripts
NM_020946.1 XM_005252109.2 XM_005252111.4 XM_006717195.3 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_017014947.1 XM_017014948.1 XM_017014949.1 XM_017014950.1 XM_017014951.1 XM_017014952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
145309 c.1786C>T p.L596F Chr.9: 123383705-123383705 on Build GRCh38
4382998 c.1672C>A p.P558T Chr.9: 123383852-123383852 on Build GRCh38
3847598 c.1746C>T p.A582A Chr.9: 123383745-123383745 on Build GRCh38
4421547 c.1780G>A p.A594T Chr.9: 123383744-123383744 on Build GRCh38
6477001 c.1854G>A p.G618G Chr.9: 123383670-123383670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866005704 Chr.9: 123383724-123383724 on Build GRCh38
rs749672773 Chr.9: 123383676-123383676 on Build GRCh38
rs779644873 Chr.9: 123383747-123383747 on Build GRCh38
rs754580261 Chr.9: 123383828-123383828 on Build GRCh38
rs201498321 Chr.9: 123383756-123383756 on Build GRCh38
rs764108452 Chr.9: 123383727-123383727 on Build GRCh38
rs770362481 Chr.9: 123383656-123383656 on Build GRCh38
rs774779902 Chr.9: 123383682-123383682 on Build GRCh38
rs771612217 Chr.9: 123383679-123383679 on Build GRCh38
rs201017866 Chr.9: 123383702-123383702 on Build GRCh38
rs200564841 Chr.9: 123383772-123383772 on Build GRCh38
rs768614573 Chr.9: 123383799-123383799 on Build GRCh38
rs778090960 Chr.9: 123383659-123383659 on Build GRCh38
rs772453295 Chr.9: 123383784-123383784 on Build GRCh38
rs371253977 Chr.9: 123383819-123383819 on Build GRCh38
rs773025069 Chr.9: 123383806-123383806 on Build GRCh38
rs751102366 Chr.9: 123383826-123383826 on Build GRCh38
rs759819093 Chr.9: 123383693-123383693 on Build GRCh38
rs1032943025 Chr.9: 123383761-123383761 on Build GRCh38
rs761735480 Chr.9: 123383734-123383734 on Build GRCh38
rs546579684 Chr.9: 123383836-123383836 on Build GRCh38
rs774855206 Chr.9: 123383717-123383717 on Build GRCh38
rs1000385483 Chr.9: 123383730-123383730 on Build GRCh38
rs199648302 Chr.9: 123383801-123383801 on Build GRCh38
rs974474775 Chr.9: 123383821-123383821 on Build GRCh38
rs749751775 Chr.9: 123383757-123383757 on Build GRCh38
rs778344810 Chr.9: 123383753-123383753 on Build GRCh38
rs377526181 Chr.9: 123383771-123383771 on Build GRCh38
rs145330954 Chr.9: 123383823-123383823 on Build GRCh38
rs955860892 Chr.9: 123383776-123383776 on Build GRCh38
rs373545651 Chr.9: 123383762-123383762 on Build GRCh38
rs1056547041 Chr.9: 123383726-123383726 on Build GRCh38
rs868735081 Chr.9: 123383645-123383645 on Build GRCh38
rs150045647 Chr.9: 123383677-123383677 on Build GRCh38
rs746131147 Chr.9: 123383780-123383780 on Build GRCh38
rs764931282 Chr.9: 123383743-123383743 on Build GRCh38
rs567551281 Chr.9: 123383745-123383745 on Build GRCh38
rs369779021 Chr.9: 123383744-123383744 on Build GRCh38
rs375269403 Chr.9: 123383696-123383696 on Build GRCh38
rs751089227 Chr.9: 123383749-123383749 on Build GRCh38
rs775679517 Chr.9: 123383701-123383701 on Build GRCh38
rs753651903 Chr.9: 123383731-123383731 on Build GRCh38
rs904658715 Chr.9: 123383690-123383690 on Build GRCh38
rs754385424 Chr.9: 123383750-123383750 on Build GRCh38
rs760816530 Chr.9: 123383790-123383790 on Build GRCh38
rs1049322400 Chr.9: 123383667-123383667 on Build GRCh38
rs754417395 Chr.9: 123383837-123383837 on Build GRCh38
rs571858526 Chr.9: 123383786-123383786 on Build GRCh38
rs776689069 Chr.9: 123383800-123383800 on Build GRCh38

View Full Product Details