Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36611885-36612158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD3
Entrez Gene ID
80206
Gene Name
formin homology 2 domain containing 3
Gene Aliases
FHOS2, Formactin2
UniGene
Hs.436636
Gene Chromosome Location
Chr.18: 36297696-36780220 on Build GRCh38
Transcripts
NM_001281739.2 NM_001281740.2 NM_025135.4 XM_005258349.1 XM_005258354.1 XM_005258355.1 XM_011526189.1 XM_011526190.1 XM_011526192.1 XM_011526193.2 XM_011526195.1 XM_011526196.1 XM_011526197.1 XM_017026006.1 XM_017026007.1 XM_017026008.1 XM_017026009.1 XM_017026010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455789 c.827T>C p.L276P Chr.18: 36611965-36611965 on Build GRCh38
2811446 c.853G>C p.V285L Chr.18: 36611991-36611991 on Build GRCh38
7368273 c.863G>A p.C288Y Chr.18: 36612001-36612001 on Build GRCh38
473807 c.913A>T p.K305* Chr.18: 36612051-36612051 on Build GRCh38
217151 c.957G>T p.E319D Chr.18: 36612095-36612095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201937289 Chr.18: 36612119-36612119 on Build GRCh38
rs769007828 Chr.18: 36611916-36611916 on Build GRCh38
rs755152519 Chr.18: 36612125-36612125 on Build GRCh38
rs760231456 Chr.18: 36611961-36611961 on Build GRCh38
rs751568896 Chr.18: 36611994-36611994 on Build GRCh38
rs766036363 Chr.18: 36611969-36611969 on Build GRCh38
rs748450224 Chr.18: 36612131-36612131 on Build GRCh38
rs370638263 Chr.18: 36612027-36612027 on Build GRCh38
rs755636461 Chr.18: 36611990-36611990 on Build GRCh38
rs894361481 Chr.18: 36611960-36611960 on Build GRCh38
rs780177528 Chr.18: 36612022-36612022 on Build GRCh38
rs751450028 Chr.18: 36612111-36612111 on Build GRCh38
rs749572966 Chr.18: 36611908-36611908 on Build GRCh38
rs555390590 Chr.18: 36611948-36611948 on Build GRCh38
rs768881340 Chr.18: 36612041-36612041 on Build GRCh38
rs916381718 Chr.18: 36612038-36612038 on Build GRCh38
rs770559364 Chr.18: 36612066-36612066 on Build GRCh38
rs762637024 Chr.18: 36612104-36612104 on Build GRCh38
rs372746494 Chr.18: 36611946-36611946 on Build GRCh38
rs61735990 Chr.18: 36612056-36612056 on Build GRCh38
rs764012361 Chr.18: 36612109-36612109 on Build GRCh38
rs199989606 Chr.18: 36612020-36612020 on Build GRCh38
rs374295739 Chr.18: 36612048-36612048 on Build GRCh38
rs978486483 Chr.18: 36611964-36611964 on Build GRCh38
rs764814944 Chr.18: 36612082-36612082 on Build GRCh38
rs533100172 Chr.18: 36612085-36612085 on Build GRCh38
rs759204801 Chr.18: 36612081-36612081 on Build GRCh38
rs748004329 Chr.18: 36611914-36611914 on Build GRCh38
rs748493835 Chr.18: 36612055-36612055 on Build GRCh38
rs773570148 Chr.18: 36612058-36612058 on Build GRCh38
rs139700928 Chr.18: 36611993-36611993 on Build GRCh38
rs924000303 Chr.18: 36611972-36611972 on Build GRCh38
rs545198325 Chr.18: 36612106-36612106 on Build GRCh38
rs377180775 Chr.18: 36611989-36611989 on Build GRCh38
rs758454008 Chr.18: 36612058-36612099 on Build GRCh38
rs370792506 Chr.18: 36611927-36611927 on Build GRCh38
rs772478823 Chr.18: 36611953-36611953 on Build GRCh38
rs146804885 Chr.18: 36611988-36611988 on Build GRCh38
rs772739269 Chr.18: 36611957-36611957 on Build GRCh38
rs986961101 Chr.18: 36612077-36612077 on Build GRCh38
rs906256113 Chr.18: 36612099-36612099 on Build GRCh38
rs776282960 Chr.18: 36612074-36612074 on Build GRCh38
rs761319775 Chr.18: 36612062-36612062 on Build GRCh38
rs759327678 Chr.18: 36611987-36611987 on Build GRCh38
rs200811037 Chr.18: 36611991-36611991 on Build GRCh38
rs753526587 Chr.18: 36611978-36611978 on Build GRCh38
rs548926459 Chr.18: 36612133-36612133 on Build GRCh38
rs73948089 Chr.18: 36612124-36612124 on Build GRCh38
rs978135845 Chr.18: 36611995-36611995 on Build GRCh38
rs778972185 Chr.18: 36612129-36612129 on Build GRCh38

View Full Product Details