Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89583676-89583923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1 XM_017023141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4341489 c.611C>T p.P204L Chr.16: 89583725-89583725 on Build GRCh38
4454843 c.593A>G p.Q198R Chr.16: 89583707-89583707 on Build GRCh38
3988731 c.585T>A p.I195I Chr.16: 89583699-89583699 on Build GRCh38
7255595 c.587delT p.V196fs*6 Chr.16: 89583701-89583701 on Build GRCh38
4652807 c.612G>A p.P204P Chr.16: 89583726-89583726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768130590 Chr.16: 89583806-89583806 on Build GRCh38
rs145568288 Chr.16: 89583752-89583752 on Build GRCh38
rs897542705 Chr.16: 89583846-89583846 on Build GRCh38
rs979585229 Chr.16: 89583897-89583898 on Build GRCh38
rs770545030 Chr.16: 89583864-89583864 on Build GRCh38
rs762806593 Chr.16: 89583823-89583823 on Build GRCh38
rs368893675 Chr.16: 89583822-89583822 on Build GRCh38
rs761407667 Chr.16: 89583764-89583764 on Build GRCh38
rs760774452 Chr.16: 89583884-89583884 on Build GRCh38
rs771257559 Chr.16: 89583799-89583799 on Build GRCh38
rs763256835 Chr.16: 89583725-89583725 on Build GRCh38
rs964595049 Chr.16: 89583885-89583885 on Build GRCh38
rs780027856 Chr.16: 89583781-89583781 on Build GRCh38
rs755079585 Chr.16: 89583785-89583785 on Build GRCh38
rs375428392 Chr.16: 89583786-89583786 on Build GRCh38
rs925366874 Chr.16: 89583899-89583899 on Build GRCh38
rs201835948 Chr.16: 89583726-89583726 on Build GRCh38
rs371647265 Chr.16: 89583732-89583732 on Build GRCh38
rs748422849 Chr.16: 89583722-89583722 on Build GRCh38
rs770099552 Chr.16: 89583790-89583790 on Build GRCh38
rs993132802 Chr.16: 89583847-89583847 on Build GRCh38
rs186909019 Chr.16: 89583901-89583901 on Build GRCh38
rs925355633 Chr.16: 89583783-89583783 on Build GRCh38
rs879861161 Chr.16: 89583839-89583839 on Build GRCh38
rs1013816422 Chr.16: 89583714-89583714 on Build GRCh38
rs896754386 Chr.16: 89583707-89583707 on Build GRCh38
rs770263088 Chr.16: 89583723-89583723 on Build GRCh38
rs1029926421 Chr.16: 89583856-89583856 on Build GRCh38
rs774730044 Chr.16: 89583740-89583740 on Build GRCh38
rs761318109 Chr.16: 89583818-89583818 on Build GRCh38
rs955171010 Chr.16: 89583810-89583810 on Build GRCh38
rs925064733 Chr.16: 89583860-89583860 on Build GRCh38
rs942865544 Chr.16: 89583711-89583711 on Build GRCh38
rs748259218 Chr.16: 89583787-89583787 on Build GRCh38
rs775620919 Chr.16: 89583801-89583801 on Build GRCh38
rs776989532 Chr.16: 89583721-89583721 on Build GRCh38
rs945060968 Chr.16: 89583830-89583830 on Build GRCh38
rs200302953 Chr.16: 89583780-89583780 on Build GRCh38
rs149729126 Chr.16: 89583719-89583719 on Build GRCh38
rs534942196 Chr.16: 89583839-89583839 on Build GRCh38
rs776080905 Chr.16: 89583812-89583812 on Build GRCh38
rs751614658 Chr.16: 89583784-89583784 on Build GRCh38
rs182566268 Chr.16: 89583797-89583797 on Build GRCh38
rs750169106 Chr.16: 89583776-89583776 on Build GRCh38
rs760161077 Chr.16: 89583750-89583750 on Build GRCh38
rs764886872 Chr.16: 89583819-89583819 on Build GRCh38
rs567442000 Chr.16: 89583838-89583838 on Build GRCh38
rs1018515798 Chr.16: 89583881-89583881 on Build GRCh38
rs779864662 Chr.16: 89583699-89583699 on Build GRCh38
rs534559082 Chr.16: 89583861-89583861 on Build GRCh38
rs375506546 Chr.16: 89583803-89583803 on Build GRCh38
rs758319422 Chr.16: 89583698-89583698 on Build GRCh38
rs1016354033 Chr.16: 89583788-89583788 on Build GRCh38
rs764831303 Chr.16: 89583770-89583770 on Build GRCh38

View Full Product Details