Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47587646-47587911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM180B
Entrez Gene ID
399888
Gene Name
family with sequence similarity 180 member B
Gene Aliases
-
UniGene
Hs.502529
Gene Chromosome Location
Chr.11: 47586678-47589194 on Build GRCh38
Transcripts
NM_001164379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6240081 c.263C>T p.P88L Chr.11: 47587805-47587805 on Build GRCh38
1585796 c.231C>T p.S77S Chr.11: 47587773-47587773 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs920816555 Chr.11: 47587780-47587780 on Build GRCh38
rs567391483 Chr.11: 47587800-47587800 on Build GRCh38
rs546647243 Chr.11: 47587806-47587806 on Build GRCh38
rs967033647 Chr.11: 47587737-47587737 on Build GRCh38
rs1023376564 Chr.11: 47587692-47587692 on Build GRCh38
rs368745483 Chr.11: 47587795-47587795 on Build GRCh38
rs1012534482 Chr.11: 47587681-47587681 on Build GRCh38
rs368443858 Chr.11: 47587754-47587754 on Build GRCh38
rs773929576 Chr.11: 47587773-47587773 on Build GRCh38
rs567086871 Chr.11: 47587819-47587819 on Build GRCh38
rs1010707974 Chr.11: 47587872-47587872 on Build GRCh38
rs754794479 Chr.11: 47587767-47587767 on Build GRCh38
rs538272330 Chr.11: 47587867-47587867 on Build GRCh38
rs906193622 Chr.11: 47587802-47587802 on Build GRCh38
rs535945744 Chr.11: 47587805-47587805 on Build GRCh38
rs554644844 Chr.11: 47587889-47587889 on Build GRCh38
rs934047547 Chr.11: 47587848-47587848 on Build GRCh38
rs764412899 Chr.11: 47587855-47587855 on Build GRCh38
rs1022190546 Chr.11: 47587699-47587699 on Build GRCh38
rs768673064 Chr.11: 47587857-47587857 on Build GRCh38
rs1022558915 Chr.11: 47587879-47587879 on Build GRCh38
rs758300997 Chr.11: 47587739-47587739 on Build GRCh38
rs1038293184 Chr.11: 47587774-47587774 on Build GRCh38
rs575997804 Chr.11: 47587854-47587854 on Build GRCh38
rs924599028 Chr.11: 47587748-47587748 on Build GRCh38
rs758659414 Chr.11: 47587806-47587806 on Build GRCh38
rs141304714 Chr.11: 47587769-47587769 on Build GRCh38
rs910171799 Chr.11: 47587801-47587801 on Build GRCh38
rs78127409 Chr.11: 47587845-47587845 on Build GRCh38
rs551269039 Chr.11: 47587750-47587750 on Build GRCh38
rs749520652 Chr.11: 47587781-47587781 on Build GRCh38
rs761076997 Chr.11: 47587847-47587847 on Build GRCh38
rs770918669 Chr.11: 47587794-47587794 on Build GRCh38
rs539196776 Chr.11: 47587821-47587821 on Build GRCh38
rs372385787 Chr.11: 47587760-47587760 on Build GRCh38
rs573408568 Chr.11: 47587785-47587785 on Build GRCh38
rs943509966 Chr.11: 47587772-47587772 on Build GRCh38

View Full Product Details