Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 80419218-80419491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA2
Entrez Gene ID
1496
Gene Name
catenin alpha 2
Gene Aliases
CAP-R, CAPR, CT114, CTNR
UniGene
Hs.167368
Gene Chromosome Location
Chr.2: 79512934-80648868 on Build GRCh38
Transcripts
NM_001164883.1 NM_001282597.2 NM_001282598.1 NM_001282599.1 NM_001282600.1 NM_001320810.1 NM_004389.3 XM_011532555.2 XM_011532556.2 XM_017003403.1 XM_017003404.1 XM_017003405.1 XM_017003406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3583598 c.1261C>T p.H421Y Chr.2: 80419470-80419470 on Build GRCh38
5901121 c.1180G>A p.E394K Chr.2: 80419491-80419491 on Build GRCh38
3583601 c.1277T>A p.F426Y Chr.2: 80419486-80419486 on Build GRCh38
4095961 c.1170C>T p.D390D Chr.2: 80419481-80419481 on Build GRCh38
5354634 c.1278C>T p.F426F Chr.2: 80419487-80419487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545104986 Chr.2: 80419461-80419461 on Build GRCh38
rs766627834 Chr.2: 80419452-80419452 on Build GRCh38
rs1052952181 Chr.2: 80419343-80419343 on Build GRCh38
rs910789445 Chr.2: 80419269-80419269 on Build GRCh38
rs767573047 Chr.2: 80419440-80419440 on Build GRCh38
rs780972183 Chr.2: 80419314-80419314 on Build GRCh38
rs745701335 Chr.2: 80419408-80419408 on Build GRCh38
rs892997691 Chr.2: 80419377-80419377 on Build GRCh38
rs770765748 Chr.2: 80419402-80419402 on Build GRCh38
rs1044353328 Chr.2: 80419392-80419392 on Build GRCh38
rs199696511 Chr.2: 80419418-80419418 on Build GRCh38
rs891736601 Chr.2: 80419361-80419361 on Build GRCh38
rs17019173 Chr.2: 80419302-80419302 on Build GRCh38
rs776568047 Chr.2: 80419407-80419407 on Build GRCh38
rs1022079608 Chr.2: 80419409-80419409 on Build GRCh38
rs773219963 Chr.2: 80419443-80419443 on Build GRCh38
rs778670332 Chr.2: 80419431-80419431 on Build GRCh38
rs1002673563 Chr.2: 80419337-80419337 on Build GRCh38
rs948557750 Chr.2: 80419391-80419391 on Build GRCh38
rs536244156 Chr.2: 80419292-80419292 on Build GRCh38
rs907021418 Chr.2: 80419306-80419306 on Build GRCh38
rs1007346770 Chr.2: 80419421-80419421 on Build GRCh38
rs757126697 Chr.2: 80419430-80419430 on Build GRCh38
rs934703889 Chr.2: 80419349-80419349 on Build GRCh38
rs746616310 Chr.2: 80419399-80419399 on Build GRCh38
rs986441615 Chr.2: 80419243-80419243 on Build GRCh38
rs759844097 Chr.2: 80419465-80419465 on Build GRCh38
rs1011575749 Chr.2: 80419372-80419372 on Build GRCh38
rs965510713 Chr.2: 80419462-80419462 on Build GRCh38
rs762802223 Chr.2: 80419436-80419436 on Build GRCh38
rs570552079 Chr.2: 80419427-80419427 on Build GRCh38
rs200870374 Chr.2: 80419401-80419401 on Build GRCh38
rs948438651 Chr.2: 80419280-80419280 on Build GRCh38

View Full Product Details