Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 17298059-17298306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTLN
Entrez Gene ID
54875
Gene Name
centlein
Gene Aliases
C9orf101, C9orf39, bA340N12.1
UniGene
Hs.435381
Gene Chromosome Location
Chr.9: 17134991-17510019 on Build GRCh38
Transcripts
NM_001114395.2 NM_017738.3 XM_005251492.1 XM_006716793.3 XM_011517941.2 XM_017014839.1 XM_017014840.1 XM_017014841.1 XM_017014842.1 XM_017014843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5089192 c.1082A>G p.D361G Chr.9: 17298288-17298288 on Build GRCh38
7210241 c.988delG p.E330fs*22 Chr.9: 17298194-17298194 on Build GRCh38
6115231 c.988G>C p.E330Q Chr.9: 17298194-17298194 on Build GRCh38
5089193 c.1082A>G p.D361G Chr.9: 17298288-17298288 on Build GRCh38
4568287 c.1070T>C p.V357A Chr.9: 17298276-17298276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746290514 Chr.9: 17298096-17298096 on Build GRCh38
rs748532437 Chr.9: 17298221-17298221 on Build GRCh38
rs920695637 Chr.9: 17298200-17298200 on Build GRCh38
rs756729000 Chr.9: 17298192-17298192 on Build GRCh38
rs910410070 Chr.9: 17298173-17298173 on Build GRCh38
rs1049831896 Chr.9: 17298156-17298156 on Build GRCh38
rs369012660 Chr.9: 17298231-17298231 on Build GRCh38
rs758462017 Chr.9: 17298180-17298180 on Build GRCh38
rs754806899 Chr.9: 17298172-17298172 on Build GRCh38
rs1047736994 Chr.9: 17298240-17298240 on Build GRCh38
rs375304931 Chr.9: 17298179-17298179 on Build GRCh38
rs954625066 Chr.9: 17298126-17298126 on Build GRCh38
rs760751354 Chr.9: 17298245-17298245 on Build GRCh38
rs573874969 Chr.9: 17298274-17298274 on Build GRCh38
rs746460015 Chr.9: 17298187-17298187 on Build GRCh38
rs759881956 Chr.9: 17298275-17298275 on Build GRCh38
rs945996507 Chr.9: 17298114-17298114 on Build GRCh38
rs373001973 Chr.9: 17298242-17298242 on Build GRCh38
rs773291417 Chr.9: 17298149-17298149 on Build GRCh38
rs1043257448 Chr.9: 17298120-17298120 on Build GRCh38
rs553820922 Chr.9: 17298095-17298095 on Build GRCh38
rs765194524 Chr.9: 17298177-17298177 on Build GRCh38
rs181389046 Chr.9: 17298130-17298130 on Build GRCh38
rs889913402 Chr.9: 17298206-17298206 on Build GRCh38
rs201918439 Chr.9: 17298268-17298268 on Build GRCh38
rs1038225087 Chr.9: 17298185-17298185 on Build GRCh38
rs572124240 Chr.9: 17298116-17298116 on Build GRCh38
rs774745524 Chr.9: 17298215-17298215 on Build GRCh38
rs776912140 Chr.9: 17298251-17298251 on Build GRCh38
rs745583204 Chr.9: 17298195-17298195 on Build GRCh38
rs766557361 Chr.9: 17298158-17298158 on Build GRCh38
rs994412800 Chr.9: 17298239-17298239 on Build GRCh38
rs987810151 Chr.9: 17298099-17298099 on Build GRCh38
rs754021280 Chr.9: 17298170-17298170 on Build GRCh38
rs780881057 Chr.9: 17298193-17298193 on Build GRCh38
rs561677677 Chr.9: 17298208-17298208 on Build GRCh38
rs766427563 Chr.9: 17298246-17298246 on Build GRCh38
rs777597179 Chr.9: 17298186-17298186 on Build GRCh38
rs773598770 Chr.9: 17298232-17298232 on Build GRCh38
rs1027677716 Chr.9: 17298250-17298250 on Build GRCh38
rs760701455 Chr.9: 17298154-17298154 on Build GRCh38
rs1008192244 Chr.9: 17298213-17298213 on Build GRCh38
rs913796637 Chr.9: 17298106-17298106 on Build GRCh38

View Full Product Details