Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23084967-23085192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD93
Entrez Gene ID
22918
Gene Name
CD93 molecule
Gene Aliases
C1QR1, C1qR(P), C1qRP, CDw93, ECSM3, MXRA4, dJ737E23.1
UniGene
Hs.97199
Gene Chromosome Location
Chr.20: 23076554-23086340 on Build GRCh38
Transcripts
NM_012072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5013572 c.1114G>A p.G372S Chr.20: 23085079-23085079 on Build GRCh38
4520102 c.1042G>A p.E348K Chr.20: 23085151-23085151 on Build GRCh38
1483494 c.1066C>T p.Q356* Chr.20: 23085127-23085127 on Build GRCh38
3544701 c.1088G>A p.G363E Chr.20: 23085105-23085105 on Build GRCh38
402200 c.1085C>A p.P362H Chr.20: 23085108-23085108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975111862 Chr.20: 23085083-23085083 on Build GRCh38
rs749061410 Chr.20: 23085097-23085097 on Build GRCh38
rs753196399 Chr.20: 23085000-23085000 on Build GRCh38
rs371193588 Chr.20: 23085066-23085066 on Build GRCh38
rs746085953 Chr.20: 23085016-23085016 on Build GRCh38
rs200444106 Chr.20: 23085101-23085101 on Build GRCh38
rs1005084781 Chr.20: 23085049-23085049 on Build GRCh38
rs142949670 Chr.20: 23085035-23085035 on Build GRCh38
rs748038221 Chr.20: 23085102-23085102 on Build GRCh38
rs772576428 Chr.20: 23085134-23085136 on Build GRCh38
rs368417861 Chr.20: 23085158-23085158 on Build GRCh38
rs150829026 Chr.20: 23085110-23085110 on Build GRCh38
rs951258683 Chr.20: 23085063-23085063 on Build GRCh38
rs767003550 Chr.20: 23085064-23085064 on Build GRCh38
rs753142971 Chr.20: 23085068-23085068 on Build GRCh38
rs139005135 Chr.20: 23085069-23085069 on Build GRCh38
rs758995480 Chr.20: 23085002-23085002 on Build GRCh38
rs1055088849 Chr.20: 23084990-23084990 on Build GRCh38
rs142218043 Chr.20: 23085096-23085096 on Build GRCh38
rs267605854 Chr.20: 23085138-23085138 on Build GRCh38
rs201414606 Chr.20: 23085092-23085092 on Build GRCh38
rs760364231 Chr.20: 23085129-23085129 on Build GRCh38
rs764669341 Chr.20: 23085071-23085071 on Build GRCh38
rs541179008 Chr.20: 23085053-23085053 on Build GRCh38
rs1046890888 Chr.20: 23085012-23085012 on Build GRCh38
rs377684251 Chr.20: 23085015-23085015 on Build GRCh38
rs772853677 Chr.20: 23085065-23085065 on Build GRCh38
rs776468245 Chr.20: 23085141-23085141 on Build GRCh38
rs757350790 Chr.20: 23085091-23085091 on Build GRCh38
rs267605855 Chr.20: 23085160-23085160 on Build GRCh38
rs879796766 Chr.20: 23085089-23085089 on Build GRCh38
rs899074925 Chr.20: 23085056-23085056 on Build GRCh38
rs146164571 Chr.20: 23085052-23085052 on Build GRCh38
rs562655027 Chr.20: 23085143-23085143 on Build GRCh38
rs963955678 Chr.20: 23085073-23085073 on Build GRCh38
rs769523197 Chr.20: 23085021-23085021 on Build GRCh38
rs555527424 Chr.20: 23084997-23084997 on Build GRCh38
rs966909806 Chr.20: 23085124-23085124 on Build GRCh38
rs373112634 Chr.20: 23085011-23085011 on Build GRCh38
rs752062112 Chr.20: 23085074-23085074 on Build GRCh38
rs747076730 Chr.20: 23085005-23085005 on Build GRCh38
rs768550295 Chr.20: 23085050-23085050 on Build GRCh38
rs767597760 Chr.20: 23085162-23085162 on Build GRCh38
rs775289910 Chr.20: 23085034-23085034 on Build GRCh38
rs750475932 Chr.20: 23085173-23085173 on Build GRCh38
rs777956213 Chr.20: 23085004-23085004 on Build GRCh38
rs760689943 Chr.20: 23085126-23085126 on Build GRCh38
rs779423494 Chr.20: 23084993-23084993 on Build GRCh38
rs764430924 Chr.20: 23085157-23085157 on Build GRCh38
rs772619730 Chr.20: 23085122-23085122 on Build GRCh38

View Full Product Details