Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78578942-78579215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA3
Entrez Gene ID
306
Gene Name
annexin A3
Gene Aliases
ANX3
UniGene
Hs.480042
Gene Chromosome Location
Chr.4: 78551588-78610451 on Build GRCh38
Transcripts
NM_005139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457884 c.166A>T p.I56F Chr.4: 78579089-78579089 on Build GRCh38
5526724 c.104G>A p.G35E Chr.4: 78579027-78579027 on Build GRCh38
4819797 c.112G>A p.E38K Chr.4: 78579035-78579035 on Build GRCh38
6020144 c.139G>C p.E47Q Chr.4: 78579062-78579062 on Build GRCh38
1057952 c.157C>T p.R53W Chr.4: 78579080-78579080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916472950 Chr.4: 78579051-78579051 on Build GRCh38
rs775883378 Chr.4: 78579007-78579007 on Build GRCh38
rs778654119 Chr.4: 78578977-78578977 on Build GRCh38
rs757000090 Chr.4: 78579170-78579170 on Build GRCh38
rs750201728 Chr.4: 78579033-78579033 on Build GRCh38
rs764161673 Chr.4: 78579015-78579015 on Build GRCh38
rs763444310 Chr.4: 78579106-78579106 on Build GRCh38
rs755896896 Chr.4: 78579061-78579061 on Build GRCh38
rs746644181 Chr.4: 78579168-78579168 on Build GRCh38
rs765302854 Chr.4: 78579156-78579156 on Build GRCh38
rs777317946 Chr.4: 78579084-78579084 on Build GRCh38
rs754940706 Chr.4: 78579153-78579153 on Build GRCh38
rs920331696 Chr.4: 78579176-78579176 on Build GRCh38
rs757867419 Chr.4: 78579159-78579159 on Build GRCh38
rs768307138 Chr.4: 78578976-78578976 on Build GRCh38
rs548941958 Chr.4: 78578997-78578997 on Build GRCh38
rs762010838 Chr.4: 78579029-78579029 on Build GRCh38
rs761922845 Chr.4: 78579127-78579127 on Build GRCh38
rs778550106 Chr.4: 78579068-78579068 on Build GRCh38
rs556360272 Chr.4: 78579175-78579175 on Build GRCh38
rs753843147 Chr.4: 78579063-78579063 on Build GRCh38
rs769212224 Chr.4: 78579112-78579112 on Build GRCh38
rs186109560 Chr.4: 78579191-78579191 on Build GRCh38
rs777425832 Chr.4: 78579166-78579166 on Build GRCh38
rs7661123 Chr.4: 78579192-78579192 on Build GRCh38
rs996919824 Chr.4: 78579066-78579066 on Build GRCh38
rs1022587953 Chr.4: 78579160-78579160 on Build GRCh38
rs770067653 Chr.4: 78579092-78579092 on Build GRCh38
rs1041016688 Chr.4: 78579032-78579032 on Build GRCh38
rs746142666 Chr.4: 78578995-78578995 on Build GRCh38
rs771906441 Chr.4: 78578984-78578984 on Build GRCh38
rs766094323 Chr.4: 78579144-78579144 on Build GRCh38
rs141446480 Chr.4: 78579062-78579062 on Build GRCh38
rs201486819 Chr.4: 78579073-78579073 on Build GRCh38
rs762961293 Chr.4: 78579011-78579011 on Build GRCh38
rs372470961 Chr.4: 78578985-78578985 on Build GRCh38
rs774394327 Chr.4: 78579125-78579125 on Build GRCh38
rs750636849 Chr.4: 78579130-78579130 on Build GRCh38
rs753756532 Chr.4: 78579150-78579150 on Build GRCh38
rs181568024 Chr.4: 78579080-78579080 on Build GRCh38
rs1031631703 Chr.4: 78579117-78579117 on Build GRCh38
rs899917618 Chr.4: 78579034-78579034 on Build GRCh38
rs200633440 Chr.4: 78579027-78579027 on Build GRCh38
rs752210769 Chr.4: 78579158-78579158 on Build GRCh38
rs199651167 Chr.4: 78579031-78579031 on Build GRCh38
rs369526651 Chr.4: 78578983-78578983 on Build GRCh38
rs944072457 Chr.4: 78579005-78579005 on Build GRCh38
rs754473655 Chr.4: 78579065-78579065 on Build GRCh38
rs760464307 Chr.4: 78579139-78579139 on Build GRCh38
rs746195293 Chr.4: 78579091-78579091 on Build GRCh38
rs778360136 Chr.4: 78579002-78579002 on Build GRCh38
rs375919538 Chr.4: 78578975-78578975 on Build GRCh38
rs970230883 Chr.4: 78579171-78579171 on Build GRCh38
rs775997466 Chr.4: 78579098-78579098 on Build GRCh38
rs146728751 Chr.4: 78579081-78579081 on Build GRCh38

View Full Product Details