Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75069268-75069499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB7
Entrez Gene ID
22
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
ABC7, ASAT, Atm1p, EST140535
UniGene
Hs.370480
Gene Chromosome Location
Chr.X: 75053172-75156340 on Build GRCh38
Transcripts
NM_001271696.1 NM_001271697.1 NM_001271698.1 NM_001271699.1 NM_004299.4

SNP Detail

SNP ID Chromosome Location
rs746001426 Chr.X: 75069469-75069469 on Build GRCh38
rs770859925 Chr.X: 75069388-75069388 on Build GRCh38
rs151288786 Chr.X: 75069328-75069328 on Build GRCh38
rs746347959 Chr.X: 75069460-75069460 on Build GRCh38
rs863223864 Chr.X: 75069334-75069334 on Build GRCh38
rs778546799 Chr.X: 75069348-75069348 on Build GRCh38
rs770382975 Chr.X: 75069473-75069473 on Build GRCh38
rs758576844 Chr.X: 75069420-75069420 on Build GRCh38
rs775967958 Chr.X: 75069479-75069479 on Build GRCh38
rs863223868 Chr.X: 75069340-75069340 on Build GRCh38
rs770922311 Chr.X: 75069436-75069436 on Build GRCh38
rs769901046 Chr.X: 75069468-75069469 on Build GRCh38
rs777989939 Chr.X: 75069425-75069425 on Build GRCh38
rs866157974 Chr.X: 75069319-75069319 on Build GRCh38
rs776840777 Chr.X: 75069452-75069452 on Build GRCh38
rs570911505 Chr.X: 75069395-75069395 on Build GRCh38
rs747035724 Chr.X: 75069434-75069434 on Build GRCh38

View Full Product Details