Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 13570858-13571085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN2
Entrez Gene ID
2199
Gene Name
fibulin 2
Gene Aliases
-
UniGene
Hs.198862
Gene Chromosome Location
Chr.3: 13549125-13638422 on Build GRCh38
Transcripts
NM_001004019.1 NM_001165035.1 NM_001998.2 XM_006713026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1038876 c.580G>A p.E194K Chr.3: 13570935-13570935 on Build GRCh38
6576881 c.539A>G p.N180S Chr.3: 13570894-13570894 on Build GRCh38
4439574 c.686C>T p.P229L Chr.3: 13571041-13571041 on Build GRCh38
1136948 c.638G>C p.G213A Chr.3: 13570993-13570993 on Build GRCh38
4750193 c.543C>A p.F181L Chr.3: 13570898-13570898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924459048 Chr.3: 13571040-13571040 on Build GRCh38
rs767247427 Chr.3: 13571037-13571037 on Build GRCh38
rs765973064 Chr.3: 13570950-13570950 on Build GRCh38
rs368996190 Chr.3: 13570991-13570991 on Build GRCh38
rs890131543 Chr.3: 13570900-13570900 on Build GRCh38
rs202191214 Chr.3: 13571009-13571009 on Build GRCh38
rs777743305 Chr.3: 13570972-13570972 on Build GRCh38
rs749057969 Chr.3: 13570977-13570977 on Build GRCh38
rs369891750 Chr.3: 13570935-13570935 on Build GRCh38
rs1055060182 Chr.3: 13570919-13570919 on Build GRCh38
rs111462798 Chr.3: 13570996-13570996 on Build GRCh38
rs752304774 Chr.3: 13571046-13571046 on Build GRCh38
rs766027162 Chr.3: 13570890-13570890 on Build GRCh38
rs781277209 Chr.3: 13570904-13570904 on Build GRCh38
rs865816597 Chr.3: 13571035-13571035 on Build GRCh38
rs555727680 Chr.3: 13570898-13570898 on Build GRCh38
rs752247665 Chr.3: 13570967-13570967 on Build GRCh38
rs776904962 Chr.3: 13570936-13570936 on Build GRCh38
rs928003663 Chr.3: 13571057-13571057 on Build GRCh38
rs764308242 Chr.3: 13570881-13570881 on Build GRCh38
rs772268800 Chr.3: 13570995-13570995 on Build GRCh38
rs751166659 Chr.3: 13570891-13570891 on Build GRCh38
rs111296106 Chr.3: 13570922-13570922 on Build GRCh38
rs773688519 Chr.3: 13570987-13570987 on Build GRCh38
rs200786749 Chr.3: 13570927-13570927 on Build GRCh38
rs562054262 Chr.3: 13570923-13570923 on Build GRCh38
rs762217546 Chr.3: 13570938-13570938 on Build GRCh38
rs757035039 Chr.3: 13570986-13570986 on Build GRCh38
rs775990815 Chr.3: 13570932-13570932 on Build GRCh38
rs112488952 Chr.3: 13571027-13571027 on Build GRCh38
rs751187121 Chr.3: 13570951-13570951 on Build GRCh38
rs762085540 Chr.3: 13571010-13571010 on Build GRCh38
rs761230016 Chr.3: 13570934-13570934 on Build GRCh38
rs200074635 Chr.3: 13571008-13571008 on Build GRCh38
rs749078519 Chr.3: 13570916-13570916 on Build GRCh38
rs377569245 Chr.3: 13571059-13571059 on Build GRCh38
rs748036614 Chr.3: 13570905-13570905 on Build GRCh38
rs371884786 Chr.3: 13570908-13570908 on Build GRCh38
rs745980717 Chr.3: 13570926-13570926 on Build GRCh38
rs374848366 Chr.3: 13570889-13570889 on Build GRCh38
rs371728000 Chr.3: 13570880-13570880 on Build GRCh38
rs939172693 Chr.3: 13571063-13571063 on Build GRCh38
rs368396364 Chr.3: 13570907-13570907 on Build GRCh38
rs371059687 Chr.3: 13571067-13571067 on Build GRCh38
rs199643829 Chr.3: 13570968-13570968 on Build GRCh38
rs113514375 Chr.3: 13570959-13570959 on Build GRCh38
rs1029337757 Chr.3: 13570970-13570970 on Build GRCh38
rs759325556 Chr.3: 13571028-13571028 on Build GRCh38

View Full Product Details