Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96840714-96840987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
UniGene
Hs.643430
Gene Chromosome Location
Chr.2: 96837914-96844021 on Build GRCh38
Transcripts
NM_144994.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1669151 c.312G>T p.Q104H Chr.2: 96840901-96840901 on Build GRCh38
1023997 c.339G>T p.E113D Chr.2: 96840874-96840874 on Build GRCh38
1023998 c.312G>A p.Q104Q Chr.2: 96840901-96840901 on Build GRCh38
4413345 c.406G>A p.D136N Chr.2: 96840807-96840807 on Build GRCh38
5645694 c.307T>G p.S103A Chr.2: 96840906-96840906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778276992 Chr.2: 96840745-96840745 on Build GRCh38
rs755012694 Chr.2: 96840738-96840738 on Build GRCh38
rs754853179 Chr.2: 96840814-96840814 on Build GRCh38
rs549401513 Chr.2: 96840914-96840914 on Build GRCh38
rs746600607 Chr.2: 96840843-96840843 on Build GRCh38
rs574411892 Chr.2: 96840766-96840766 on Build GRCh38
rs370266096 Chr.2: 96840954-96840954 on Build GRCh38
rs113139125 Chr.2: 96840784-96840784 on Build GRCh38
rs781751846 Chr.2: 96840870-96840879 on Build GRCh38
rs762423177 Chr.2: 96840881-96840881 on Build GRCh38
rs543361284 Chr.2: 96840769-96840769 on Build GRCh38
rs961820422 Chr.2: 96840903-96840903 on Build GRCh38
rs747709958 Chr.2: 96840752-96840752 on Build GRCh38
rs1057455029 Chr.2: 96840794-96840794 on Build GRCh38
rs749699327 Chr.2: 96840868-96840868 on Build GRCh38
rs759336163 Chr.2: 96840925-96840925 on Build GRCh38
rs374364165 Chr.2: 96840910-96840910 on Build GRCh38
rs201801059 Chr.2: 96840943-96840943 on Build GRCh38
rs939214919 Chr.2: 96840780-96840780 on Build GRCh38
rs369629521 Chr.2: 96840871-96840871 on Build GRCh38
rs751115977 Chr.2: 96840947-96840947 on Build GRCh38
rs564075025 Chr.2: 96840856-96840856 on Build GRCh38
rs758648233 Chr.2: 96840818-96840818 on Build GRCh38
rs761940682 Chr.2: 96840789-96840789 on Build GRCh38
rs535055195 Chr.2: 96840953-96840953 on Build GRCh38
rs373891608 Chr.2: 96840737-96840737 on Build GRCh38
rs752576397 Chr.2: 96840938-96840938 on Build GRCh38
rs200734536 Chr.2: 96840905-96840905 on Build GRCh38
rs748674513 Chr.2: 96840881-96840881 on Build GRCh38
rs773490106 Chr.2: 96840911-96840911 on Build GRCh38
rs560056848 Chr.2: 96840792-96840792 on Build GRCh38
rs145216473 Chr.2: 96840878-96840878 on Build GRCh38
rs377042453 Chr.2: 96840836-96840836 on Build GRCh38
rs1012077666 Chr.2: 96840832-96840832 on Build GRCh38
rs112667673 Chr.2: 96840786-96840786 on Build GRCh38
rs777280854 Chr.2: 96840837-96840837 on Build GRCh38
rs766673942 Chr.2: 96840917-96840917 on Build GRCh38
rs780257033 Chr.2: 96840861-96840861 on Build GRCh38
rs769178299 Chr.2: 96840829-96840832 on Build GRCh38
rs746686767 Chr.2: 96840765-96840765 on Build GRCh38
rs770213852 Chr.2: 96840925-96840929 on Build GRCh38
rs985217903 Chr.2: 96840957-96840957 on Build GRCh38
rs372587786 Chr.2: 96840961-96840961 on Build GRCh38
rs777262447 Chr.2: 96840946-96840946 on Build GRCh38
rs201808510 Chr.2: 96840833-96840833 on Build GRCh38
rs576366922 Chr.2: 96840741-96840741 on Build GRCh38
rs760980110 Chr.2: 96840802-96840802 on Build GRCh38
rs764968089 Chr.2: 96840931-96840931 on Build GRCh38
rs766085996 Chr.2: 96840810-96840810 on Build GRCh38
rs769253900 Chr.2: 96840783-96840783 on Build GRCh38
rs771686209 Chr.2: 96840755-96840755 on Build GRCh38
rs772811646 Chr.2: 96840764-96840764 on Build GRCh38
rs139291784 Chr.2: 96840813-96840813 on Build GRCh38
rs753546209 Chr.2: 96840919-96840919 on Build GRCh38

View Full Product Details