Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38501276-38501549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOG
Entrez Gene ID
9941
Gene Name
endo/exonuclease (5'-3'), endonuclease G-like
Gene Aliases
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB
UniGene
Hs.517897
Gene Chromosome Location
Chr.3: 38496127-38526305 on Build GRCh38
Transcripts
NM_001145464.1 NM_001318954.1 NM_001318955.1 NM_001318956.1 NM_001318957.1 NM_001318958.1 NM_001318959.1 NM_005107.3 NR_134938.1 XM_005265633.3 XM_005265635.3 XM_006713442.3 XM_006713444.3 XM_017007584.1 XM_017007585.1 XM_017007586.1 XM_017007587.1 XM_017007588.1 XM_017007589.1 XM_017007590.1 XM_017007591.1 XM_017007592.1 XM_017007593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
350865 c.449C>A p.S150* Chr.3: 38501490-38501490 on Build GRCh38
5627235 c.8C>G p.P3R Chr.3: 38501469-38501469 on Build GRCh38
4117290 c.395T>C p.V132A Chr.3: 38501436-38501436 on Build GRCh38
6047086 c.362C>A p.P121H Chr.3: 38501403-38501403 on Build GRCh38
3392376 c.383A>G p.N128S Chr.3: 38501424-38501424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761112726 Chr.3: 38501407-38501407 on Build GRCh38
rs766423067 Chr.3: 38501445-38501445 on Build GRCh38
rs566053711 Chr.3: 38501296-38501296 on Build GRCh38
rs200655025 Chr.3: 38501391-38501391 on Build GRCh38
rs778748335 Chr.3: 38501505-38501505 on Build GRCh38
rs777698117 Chr.3: 38501454-38501454 on Build GRCh38
rs775176182 Chr.3: 38501517-38501517 on Build GRCh38
rs754186302 Chr.3: 38501306-38501306 on Build GRCh38
rs748043553 Chr.3: 38501331-38501331 on Build GRCh38
rs750512965 Chr.3: 38501323-38501323 on Build GRCh38
rs923935369 Chr.3: 38501301-38501301 on Build GRCh38
rs777538199 Chr.3: 38501341-38501341 on Build GRCh38
rs746463300 Chr.3: 38501522-38501522 on Build GRCh38
rs866655619 Chr.3: 38501387-38501387 on Build GRCh38
rs150471797 Chr.3: 38501446-38501446 on Build GRCh38
rs1025500278 Chr.3: 38501338-38501338 on Build GRCh38
rs771804473 Chr.3: 38501512-38501512 on Build GRCh38
rs368035903 Chr.3: 38501326-38501326 on Build GRCh38
rs745494772 Chr.3: 38501506-38501506 on Build GRCh38
rs935287807 Chr.3: 38501317-38501317 on Build GRCh38
rs774135514 Chr.3: 38501364-38501364 on Build GRCh38
rs770955355 Chr.3: 38501390-38501390 on Build GRCh38
rs947859324 Chr.3: 38501334-38501334 on Build GRCh38
rs771677731 Chr.3: 38501393-38501393 on Build GRCh38
rs373915238 Chr.3: 38501308-38501308 on Build GRCh38
rs757116415 Chr.3: 38501494-38501494 on Build GRCh38
rs764626582 Chr.3: 38501408-38501408 on Build GRCh38
rs758510435 Chr.3: 38501435-38501435 on Build GRCh38
rs749069435 Chr.3: 38501356-38501356 on Build GRCh38
rs765498225 Chr.3: 38501418-38501418 on Build GRCh38
rs202058731 Chr.3: 38501414-38501414 on Build GRCh38
rs749029952 Chr.3: 38501335-38501335 on Build GRCh38
rs776188991 Chr.3: 38501405-38501405 on Build GRCh38
rs749096148 Chr.3: 38501493-38501493 on Build GRCh38
rs746023160 Chr.3: 38501409-38501409 on Build GRCh38
rs756034502 Chr.3: 38501453-38501453 on Build GRCh38
rs1017145502 Chr.3: 38501304-38501304 on Build GRCh38
rs781170120 Chr.3: 38501328-38501328 on Build GRCh38
rs955544107 Chr.3: 38501490-38501490 on Build GRCh38
rs765379049 Chr.3: 38501311-38501311 on Build GRCh38
rs1044836985 Chr.3: 38501369-38501369 on Build GRCh38
rs371259940 Chr.3: 38501343-38501343 on Build GRCh38
rs200753937 Chr.3: 38501424-38501424 on Build GRCh38

View Full Product Details