Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122937984-122938219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf88
Entrez Gene ID
80007
Gene Name
chromosome 10 open reading frame 88
Gene Aliases
-
UniGene
Hs.501286
Gene Chromosome Location
Chr.10: 122930903-122954403 on Build GRCh38
Transcripts
NM_024942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6249178 c.764C>A p.T255K Chr.10: 122938044-122938044 on Build GRCh38
1603098 c.812A>G p.N271S Chr.10: 122937996-122937996 on Build GRCh38
6128795 c.711G>A p.M237I Chr.10: 122938097-122938097 on Build GRCh38
3790562 c.725C>T p.S242F Chr.10: 122938083-122938083 on Build GRCh38
682943 c.739G>T p.G247* Chr.10: 122938069-122938069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969715060 Chr.10: 122938069-122938069 on Build GRCh38
rs760744664 Chr.10: 122938092-122938092 on Build GRCh38
rs777301305 Chr.10: 122938117-122938117 on Build GRCh38
rs963166669 Chr.10: 122938187-122938187 on Build GRCh38
rs746235322 Chr.10: 122938141-122938141 on Build GRCh38
rs781678327 Chr.10: 122938139-122938139 on Build GRCh38
rs757839396 Chr.10: 122938022-122938022 on Build GRCh38
rs113796676 Chr.10: 122938131-122938131 on Build GRCh38
rs376048882 Chr.10: 122938057-122938057 on Build GRCh38
rs149187662 Chr.10: 122938101-122938101 on Build GRCh38
rs775244366 Chr.10: 122938178-122938184 on Build GRCh38
rs1013484491 Chr.10: 122938088-122938088 on Build GRCh38
rs769917366 Chr.10: 122938065-122938065 on Build GRCh38
rs757931782 Chr.10: 122938116-122938116 on Build GRCh38
rs764808792 Chr.10: 122938113-122938113 on Build GRCh38
rs192724347 Chr.10: 122938053-122938053 on Build GRCh38
rs35980281 Chr.10: 122938012-122938013 on Build GRCh38
rs370547065 Chr.10: 122938178-122938178 on Build GRCh38
rs1002046181 Chr.10: 122938017-122938017 on Build GRCh38
rs1033511131 Chr.10: 122938042-122938042 on Build GRCh38
rs200060782 Chr.10: 122938085-122938085 on Build GRCh38
rs371333911 Chr.10: 122938128-122938128 on Build GRCh38
rs774542782 Chr.10: 122938089-122938089 on Build GRCh38
rs752342938 Chr.10: 122938115-122938115 on Build GRCh38
rs763150388 Chr.10: 122938070-122938070 on Build GRCh38
rs778286602 Chr.10: 122938027-122938027 on Build GRCh38
rs374582513 Chr.10: 122938193-122938193 on Build GRCh38
rs983241544 Chr.10: 122938134-122938134 on Build GRCh38
rs747469218 Chr.10: 122938043-122938043 on Build GRCh38
rs747625450 Chr.10: 122938019-122938019 on Build GRCh38
rs770150043 Chr.10: 122938165-122938165 on Build GRCh38
rs754735554 Chr.10: 122938104-122938104 on Build GRCh38
rs775727515 Chr.10: 122938066-122938066 on Build GRCh38
rs1017024525 Chr.10: 122938166-122938166 on Build GRCh38
rs766405755 Chr.10: 122938094-122938094 on Build GRCh38
rs145904265 Chr.10: 122938127-122938127 on Build GRCh38
rs771408428 Chr.10: 122938052-122938052 on Build GRCh38
rs574050228 Chr.10: 122938118-122938118 on Build GRCh38

View Full Product Details