Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70346857-70347116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19A
Entrez Gene ID
55308
Gene Name
DEAD-box helicase 19A
Gene Aliases
DDX19-DDX19L, DDX19L
UniGene
Hs.656037
Gene Chromosome Location
Chr.16: 70346829-70373383 on Build GRCh38
Transcripts
NM_001320522.1 NM_001320525.1 NM_001320526.1 NM_001320527.1 NM_018332.4 NR_135296.1

SNP Detail

SNP ID Chromosome Location
rs552866340 Chr.16: 70347064-70347064 on Build GRCh38
rs765265868 Chr.16: 70346933-70346933 on Build GRCh38
rs563254627 Chr.16: 70346893-70346893 on Build GRCh38
rs749707741 Chr.16: 70346915-70346915 on Build GRCh38
rs1010474539 Chr.16: 70346922-70346922 on Build GRCh38
rs748657054 Chr.16: 70346952-70346952 on Build GRCh38
rs759828866 Chr.16: 70347012-70347012 on Build GRCh38
rs747643245 Chr.16: 70346921-70346921 on Build GRCh38
rs550688658 Chr.16: 70346935-70346935 on Build GRCh38
rs1021019609 Chr.16: 70346926-70346926 on Build GRCh38
rs763019372 Chr.16: 70346979-70346979 on Build GRCh38
rs575348122 Chr.16: 70346894-70346894 on Build GRCh38
rs984416257 Chr.16: 70346946-70346946 on Build GRCh38
rs201672351 Chr.16: 70346971-70346971 on Build GRCh38
rs745523386 Chr.16: 70347078-70347078 on Build GRCh38
rs772797467 Chr.16: 70346882-70346882 on Build GRCh38
rs751659493 Chr.16: 70346904-70346904 on Build GRCh38
rs766661312 Chr.16: 70346941-70346941 on Build GRCh38
rs771708430 Chr.16: 70346877-70346877 on Build GRCh38
rs746670593 Chr.16: 70346930-70346930 on Build GRCh38
rs779970075 Chr.16: 70347087-70347087 on Build GRCh38
rs545113215 Chr.16: 70346883-70346883 on Build GRCh38
rs910846772 Chr.16: 70346989-70346989 on Build GRCh38
rs770460223 Chr.16: 70346964-70346964 on Build GRCh38
rs769728577 Chr.16: 70346975-70346975 on Build GRCh38
rs567407454 Chr.16: 70347000-70347000 on Build GRCh38
rs758014660 Chr.16: 70346929-70346929 on Build GRCh38
rs756265560 Chr.16: 70346936-70346936 on Build GRCh38
rs1051225342 Chr.16: 70346902-70346902 on Build GRCh38
rs751107268 Chr.16: 70347095-70347100 on Build GRCh38
rs151075279 Chr.16: 70346932-70346932 on Build GRCh38
rs775505678 Chr.16: 70346978-70346978 on Build GRCh38
rs779333848 Chr.16: 70346949-70346949 on Build GRCh38
rs962605259 Chr.16: 70346977-70346977 on Build GRCh38
rs190797951 Chr.16: 70346924-70346924 on Build GRCh38
rs528783026 Chr.16: 70346919-70346919 on Build GRCh38
rs561613807 Chr.16: 70346910-70346910 on Build GRCh38
rs773132234 Chr.16: 70346998-70346998 on Build GRCh38
rs772853693 Chr.16: 70346929-70346929 on Build GRCh38
rs568894745 Chr.16: 70346942-70346942 on Build GRCh38
rs775630463 Chr.16: 70346934-70346934 on Build GRCh38
rs45472097 Chr.16: 70347042-70347042 on Build GRCh38
rs376800227 Chr.16: 70347088-70347088 on Build GRCh38
rs536295255 Chr.16: 70346968-70346968 on Build GRCh38
rs143242388 Chr.16: 70346967-70346968 on Build GRCh38
rs746579803 Chr.16: 70346960-70346960 on Build GRCh38
rs373843943 Chr.16: 70346973-70346973 on Build GRCh38
rs765315208 Chr.16: 70346900-70346900 on Build GRCh38
rs757883952 Chr.16: 70346954-70346954 on Build GRCh38
rs751028803 Chr.16: 70347059-70347059 on Build GRCh38
rs756853029 Chr.16: 70347061-70347061 on Build GRCh38
rs987210832 Chr.16: 70347069-70347069 on Build GRCh38
rs777172647 Chr.16: 70346959-70346959 on Build GRCh38
rs771583821 Chr.16: 70346923-70346923 on Build GRCh38
rs768469452 Chr.16: 70347094-70347094 on Build GRCh38
rs765618719 Chr.16: 70347025-70347025 on Build GRCh38
rs565490385 Chr.16: 70346931-70346931 on Build GRCh38
rs933975322 Chr.16: 70347062-70347062 on Build GRCh38
rs753152601 Chr.16: 70347038-70347038 on Build GRCh38
rs753965388 Chr.16: 70346881-70346881 on Build GRCh38
rs764138187 Chr.16: 70346987-70346987 on Build GRCh38
rs756436962 Chr.16: 70346906-70346906 on Build GRCh38
rs1065158 Chr.16: 70347006-70347006 on Build GRCh38
rs943649379 Chr.16: 70347037-70347037 on Build GRCh38
rs961222188 Chr.16: 70346957-70346957 on Build GRCh38
rs755393414 Chr.16: 70346947-70346947 on Build GRCh38
rs766493362 Chr.16: 70347003-70347003 on Build GRCh38
rs548182317 Chr.16: 70346974-70346974 on Build GRCh38
rs757359637 Chr.16: 70346892-70346892 on Build GRCh38
rs755442763 Chr.16: 70346916-70346916 on Build GRCh38
rs767771170 Chr.16: 70346905-70346905 on Build GRCh38
rs543332759 Chr.16: 70346899-70346899 on Build GRCh38

View Full Product Details