Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18573851-18574110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
UniGene
Hs.741483
Gene Chromosome Location
Chr.9: 17906574-18910950 on Build GRCh38
Transcripts
NM_001040272.5 NM_052866.4 XM_011518063.2 XM_011518064.2 XM_017015310.1 XM_017015311.1 XM_017015312.1 XM_017015313.1 XM_017015314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3848201 c.266G>A p.R89Q Chr.9: 18574058-18574058 on Build GRCh38
5226983 c.257G>A p.G86D Chr.9: 18574049-18574049 on Build GRCh38
3656384 c.283G>A p.A95T Chr.9: 18574075-18574075 on Build GRCh38
7085509 c.310C>A p.Q104K Chr.9: 18574102-18574102 on Build GRCh38
455729 c.257G>A p.G86D Chr.9: 18574049-18574049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746920712 Chr.9: 18574086-18574086 on Build GRCh38
rs1030583059 Chr.9: 18573923-18573923 on Build GRCh38
rs908230000 Chr.9: 18573976-18573976 on Build GRCh38
rs774981030 Chr.9: 18573988-18573988 on Build GRCh38
rs758224258 Chr.9: 18574076-18574076 on Build GRCh38
rs773673361 Chr.9: 18574039-18574039 on Build GRCh38
rs762410633 Chr.9: 18573981-18573981 on Build GRCh38
rs755138687 Chr.9: 18574058-18574058 on Build GRCh38
rs777782482 Chr.9: 18573993-18573993 on Build GRCh38
rs936723388 Chr.9: 18573888-18573888 on Build GRCh38
rs761271486 Chr.9: 18574040-18574040 on Build GRCh38
rs770296961 Chr.9: 18574000-18574000 on Build GRCh38
rs746973703 Chr.9: 18573997-18573997 on Build GRCh38
rs41302063 Chr.9: 18574015-18574015 on Build GRCh38
rs889339097 Chr.9: 18573958-18573958 on Build GRCh38
rs757257913 Chr.9: 18574087-18574087 on Build GRCh38
rs372350270 Chr.9: 18574004-18574004 on Build GRCh38
rs1033400701 Chr.9: 18573952-18573952 on Build GRCh38
rs200442699 Chr.9: 18574056-18574056 on Build GRCh38
rs566073161 Chr.9: 18573948-18573948 on Build GRCh38
rs774686444 Chr.9: 18574021-18574021 on Build GRCh38
rs566217072 Chr.9: 18573983-18573983 on Build GRCh38
rs765440505 Chr.9: 18574065-18574065 on Build GRCh38
rs190050504 Chr.9: 18574073-18574073 on Build GRCh38
rs753994179 Chr.9: 18574043-18574043 on Build GRCh38
rs559061355 Chr.9: 18574013-18574013 on Build GRCh38
rs1014528889 Chr.9: 18573910-18573910 on Build GRCh38
rs146968698 Chr.9: 18573905-18573905 on Build GRCh38
rs185147701 Chr.9: 18574009-18574009 on Build GRCh38
rs759078761 Chr.9: 18574014-18574014 on Build GRCh38
rs772002375 Chr.9: 18573989-18573989 on Build GRCh38
rs764483928 Chr.9: 18574034-18574034 on Build GRCh38
rs753959806 Chr.9: 18573982-18573982 on Build GRCh38
rs576641191 Chr.9: 18573985-18573985 on Build GRCh38
rs1016819175 Chr.9: 18574002-18574002 on Build GRCh38
rs13300372 Chr.9: 18573920-18573920 on Build GRCh38
rs777733113 Chr.9: 18574081-18574081 on Build GRCh38
rs762107707 Chr.9: 18574026-18574026 on Build GRCh38
rs149465597 Chr.9: 18574038-18574038 on Build GRCh38
rs952569787 Chr.9: 18573954-18573954 on Build GRCh38
rs763690710 Chr.9: 18574017-18574018 on Build GRCh38
rs201595808 Chr.9: 18573981-18573981 on Build GRCh38
rs539093080 Chr.9: 18573914-18573914 on Build GRCh38

View Full Product Details