Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99679256-99679529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_020354.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2057404 c.307C>T p.P103S Chr.10: 99679376-99679376 on Build GRCh38
274974 c.242A>C p.N81T Chr.10: 99679311-99679311 on Build GRCh38
1345229 c.382delA p.I130fs*23 Chr.10: 99679451-99679451 on Build GRCh38
5693281 c.235G>T p.D79Y Chr.10: 99679304-99679304 on Build GRCh38
74520 c.324C>A p.N108K Chr.10: 99679393-99679393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781127847 Chr.10: 99679294-99679294 on Build GRCh38
rs762690351 Chr.10: 99679497-99679497 on Build GRCh38
rs778371833 Chr.10: 99679478-99679478 on Build GRCh38
rs368620286 Chr.10: 99679431-99679431 on Build GRCh38
rs748607179 Chr.10: 99679473-99679473 on Build GRCh38
rs763947377 Chr.10: 99679412-99679412 on Build GRCh38
rs749614688 Chr.10: 99679338-99679338 on Build GRCh38
rs562711164 Chr.10: 99679466-99679466 on Build GRCh38
rs769032311 Chr.10: 99679342-99679342 on Build GRCh38
rs774747547 Chr.10: 99679354-99679354 on Build GRCh38
rs780203334 Chr.10: 99679442-99679442 on Build GRCh38
rs764753437 Chr.10: 99679393-99679393 on Build GRCh38
rs746016879 Chr.10: 99679297-99679297 on Build GRCh38
rs754036998 Chr.10: 99679450-99679450 on Build GRCh38
rs554004708 Chr.10: 99679470-99679470 on Build GRCh38
rs1004447088 Chr.10: 99679400-99679400 on Build GRCh38
rs200759032 Chr.10: 99679364-99679364 on Build GRCh38
rs372561364 Chr.10: 99679329-99679329 on Build GRCh38
rs761642202 Chr.10: 99679417-99679417 on Build GRCh38
rs775176465 Chr.10: 99679496-99679496 on Build GRCh38
rs762739508 Chr.10: 99679405-99679405 on Build GRCh38
rs199591970 Chr.10: 99679355-99679355 on Build GRCh38
rs3740079 Chr.10: 99679495-99679495 on Build GRCh38
rs775301582 Chr.10: 99679399-99679399 on Build GRCh38
rs775993892 Chr.10: 99679356-99679356 on Build GRCh38
rs751466685 Chr.10: 99679416-99679416 on Build GRCh38
rs780260445 Chr.10: 99679323-99679323 on Build GRCh38
rs767501634 Chr.10: 99679430-99679430 on Build GRCh38
rs745313555 Chr.10: 99679488-99679488 on Build GRCh38
rs377065169 Chr.10: 99679429-99679429 on Build GRCh38
rs768294416 Chr.10: 99679377-99679381 on Build GRCh38
rs147379720 Chr.10: 99679459-99679459 on Build GRCh38
rs954032389 Chr.10: 99679483-99679483 on Build GRCh38
rs896441949 Chr.10: 99679306-99679306 on Build GRCh38
rs756231191 Chr.10: 99679298-99679298 on Build GRCh38
rs372653473 Chr.10: 99679475-99679475 on Build GRCh38
rs1015366487 Chr.10: 99679471-99679471 on Build GRCh38
rs759102720 Chr.10: 99679357-99679357 on Build GRCh38

View Full Product Details