Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159337386-159337631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
UniGene
Hs.470369
Gene Chromosome Location
Chr.2: 159315312-159712439 on Build GRCh38
Transcripts
NM_001289975.1 NM_013450.3 XM_005246488.2 XM_005246489.4 XM_005246492.4 XM_006712461.3 XM_011511038.2 XM_011511039.1 XM_011511042.1 XM_011511043.1 XM_011511044.1 XM_011511045.1 XM_011511047.1 XM_011511048.2 XM_011511050.1 XM_011511051.1 XM_011511052.1 XM_011511053.2 XM_011511054.2 XM_011511056.1 XM_017003918.1 XM_017003919.1 XM_017003920.1 XM_017003921.1 XM_017003922.1 XM_017003923.1 XM_017003924.1 XM_017003925.1 XM_017003926.1 XM_017003927.1 XM_017003928.1 XM_017003929.1 XM_017003930.1 XM_017003931.1 XM_017003932.1 XM_017003933.1 XM_017003934.1 XM_017003935.1 XM_017003936.1 XM_017003937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305744 c.5648A>T p.N1883I Chr.2: 159337579-159337579 on Build GRCh38
6610085 c.5645G>A p.R1882Q Chr.2: 159337582-159337582 on Build GRCh38
3300224 c.5655A>G p.E1885E Chr.2: 159337572-159337572 on Build GRCh38
1613551 c.5614A>G p.I1872V Chr.2: 159337613-159337613 on Build GRCh38
5764334 c.5632G>A p.A1878T Chr.2: 159337595-159337595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770844066 Chr.2: 159337528-159337528 on Build GRCh38
rs780290419 Chr.2: 159337495-159337495 on Build GRCh38
rs750801985 Chr.2: 159337492-159337492 on Build GRCh38
rs749238515 Chr.2: 159337518-159337518 on Build GRCh38
rs775234669 Chr.2: 159337551-159337551 on Build GRCh38
rs201017757 Chr.2: 159337583-159337583 on Build GRCh38
rs755106961 Chr.2: 159337503-159337503 on Build GRCh38
rs778748909 Chr.2: 159337532-159337532 on Build GRCh38
rs768213384 Chr.2: 159337576-159337576 on Build GRCh38
rs893694747 Chr.2: 159337463-159337463 on Build GRCh38
rs7559010 Chr.2: 159337481-159337481 on Build GRCh38
rs752583056 Chr.2: 159337448-159337449 on Build GRCh38
rs750857206 Chr.2: 159337602-159337602 on Build GRCh38
rs760214543 Chr.2: 159337440-159337440 on Build GRCh38
rs1029707129 Chr.2: 159337424-159337424 on Build GRCh38
rs372442188 Chr.2: 159337464-159337464 on Build GRCh38
rs200039167 Chr.2: 159337582-159337582 on Build GRCh38
rs760201416 Chr.2: 159337568-159337568 on Build GRCh38
rs763468095 Chr.2: 159337442-159337442 on Build GRCh38
rs745634469 Chr.2: 159337542-159337542 on Build GRCh38
rs935131025 Chr.2: 159337416-159337416 on Build GRCh38
rs767197066 Chr.2: 159337429-159337429 on Build GRCh38
rs201605818 Chr.2: 159337550-159337550 on Build GRCh38
rs781182745 Chr.2: 159337504-159337504 on Build GRCh38
rs539654244 Chr.2: 159337482-159337482 on Build GRCh38
rs35553195 Chr.2: 159337434-159337435 on Build GRCh38
rs765833630 Chr.2: 159337599-159337599 on Build GRCh38
rs557549279 Chr.2: 159337494-159337494 on Build GRCh38

View Full Product Details