Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36356578-36356845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIM1
Entrez Gene ID
51232
Gene Name
cysteine rich transmembrane BMP regulator 1
Gene Aliases
CRIM-1, S52
UniGene
Hs.699247
Gene Chromosome Location
Chr.2: 36356088-36551135 on Build GRCh38
Transcripts
NM_016441.2 XM_005264357.4 XM_011532898.2 XM_011532899.2 XM_011532901.2 XM_017004258.1 XM_017004259.1 XM_017004260.1 XM_017004261.1 XM_017004262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5978326 c.331+1G>T p.? Chr.2: 36356624-36356624 on Build GRCh38
6579668 c.322G>T p.V108F Chr.2: 36356614-36356614 on Build GRCh38
1284304 c.318G>C p.A106A Chr.2: 36356610-36356610 on Build GRCh38
6564722 c.301C>T p.L101F Chr.2: 36356593-36356593 on Build GRCh38
6635488 c.288C>G p.L96L Chr.2: 36356580-36356580 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760629421 Chr.2: 36356643-36356643 on Build GRCh38
rs767359122 Chr.2: 36356811-36356811 on Build GRCh38
rs528529738 Chr.2: 36356635-36356635 on Build GRCh38
rs756704237 Chr.2: 36356672-36356672 on Build GRCh38
rs748488530 Chr.2: 36356627-36356627 on Build GRCh38
rs941348822 Chr.2: 36356694-36356694 on Build GRCh38
rs554144203 Chr.2: 36356680-36356680 on Build GRCh38
rs779745197 Chr.2: 36356665-36356665 on Build GRCh38
rs1002049748 Chr.2: 36356760-36356760 on Build GRCh38
rs749410861 Chr.2: 36356633-36356633 on Build GRCh38
rs777877565 Chr.2: 36356728-36356728 on Build GRCh38
rs967365438 Chr.2: 36356793-36356793 on Build GRCh38
rs1033965682 Chr.2: 36356771-36356771 on Build GRCh38
rs565244102 Chr.2: 36356704-36356704 on Build GRCh38
rs140753944 Chr.2: 36356616-36356616 on Build GRCh38
rs899571887 Chr.2: 36356689-36356689 on Build GRCh38
rs375489342 Chr.2: 36356645-36356645 on Build GRCh38
rs1032802753 Chr.2: 36356753-36356753 on Build GRCh38
rs572362767 Chr.2: 36356720-36356720 on Build GRCh38
rs899918265 Chr.2: 36356707-36356707 on Build GRCh38
rs1051658672 Chr.2: 36356713-36356713 on Build GRCh38
rs891383511 Chr.2: 36356714-36356714 on Build GRCh38
rs778218633 Chr.2: 36356674-36356674 on Build GRCh38
rs763821741 Chr.2: 36356644-36356644 on Build GRCh38
rs569556802 Chr.2: 36356725-36356725 on Build GRCh38
rs775367541 Chr.2: 36356641-36356641 on Build GRCh38
rs113549769 Chr.2: 36356628-36356628 on Build GRCh38
rs567060175 Chr.2: 36356808-36356808 on Build GRCh38
rs371784720 Chr.2: 36356637-36356637 on Build GRCh38
rs753638084 Chr.2: 36356646-36356646 on Build GRCh38
rs773752408 Chr.2: 36356805-36356805 on Build GRCh38
rs866267256 Chr.2: 36356688-36356688 on Build GRCh38
rs958816409 Chr.2: 36356786-36356786 on Build GRCh38
rs536993116 Chr.2: 36356759-36356759 on Build GRCh38
rs999849471 Chr.2: 36356744-36356744 on Build GRCh38
rs917237335 Chr.2: 36356792-36356792 on Build GRCh38
rs114612022 Chr.2: 36356663-36356663 on Build GRCh38
rs902874317 Chr.2: 36356733-36356733 on Build GRCh38
rs749569514 Chr.2: 36356820-36356820 on Build GRCh38
rs992898677 Chr.2: 36356787-36356787 on Build GRCh38
rs1016876853 Chr.2: 36356749-36356749 on Build GRCh38
rs937307006 Chr.2: 36356619-36356619 on Build GRCh38
rs886802816 Chr.2: 36356715-36356715 on Build GRCh38
rs548496801 Chr.2: 36356803-36356803 on Build GRCh38
rs933742488 Chr.2: 36356814-36356814 on Build GRCh38
rs745453573 Chr.2: 36356609-36356609 on Build GRCh38
rs113501362 Chr.2: 36356659-36356659 on Build GRCh38
rs781775912 Chr.2: 36356620-36356620 on Build GRCh38
rs761370373 Chr.2: 36356652-36356652 on Build GRCh38
rs765001909 Chr.2: 36356653-36356653 on Build GRCh38
rs759271357 Chr.2: 36356625-36356626 on Build GRCh38
rs977620712 Chr.2: 36356799-36356799 on Build GRCh38
rs756950848 Chr.2: 36356600-36356600 on Build GRCh38
rs1050730549 Chr.2: 36356712-36356712 on Build GRCh38
rs749883664 Chr.2: 36356660-36356660 on Build GRCh38
rs1019966949 Chr.2: 36356730-36356730 on Build GRCh38
rs778222123 Chr.2: 36356601-36356601 on Build GRCh38
rs551023552 Chr.2: 36356721-36356721 on Build GRCh38
rs753208601 Chr.2: 36356671-36356671 on Build GRCh38

View Full Product Details