Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102350257-102350514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC2
Entrez Gene ID
329
Gene Name
baculoviral IAP repeat containing 2
Gene Aliases
API1, HIAP2, Hiap-2, MIHB, RNF48, c-IAP1, cIAP1
UniGene
Hs.696238
Gene Chromosome Location
Chr.11: 102347182-102378670 on Build GRCh38
Transcripts
NM_001166.4 NM_001256163.1 NM_001256166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
95580 c.605C>G p.S202* Chr.11: 102350459-102350459 on Build GRCh38
324816 c.555A>T p.E185D Chr.11: 102350409-102350409 on Build GRCh38
318925 c.638G>T p.G213V Chr.11: 102350492-102350492 on Build GRCh38
5772975 c.471T>G p.S157S Chr.11: 102350325-102350325 on Build GRCh38
5020386 c.532A>G p.S178G Chr.11: 102350386-102350386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747200705 Chr.11: 102350478-102350478 on Build GRCh38
rs757139875 Chr.11: 102350440-102350440 on Build GRCh38
rs372125025 Chr.11: 102350384-102350384 on Build GRCh38
rs751116057 Chr.11: 102350326-102350326 on Build GRCh38
rs753726394 Chr.11: 102350439-102350439 on Build GRCh38
rs991945065 Chr.11: 102350334-102350334 on Build GRCh38
rs372507082 Chr.11: 102350410-102350410 on Build GRCh38
rs762201946 Chr.11: 102350401-102350401 on Build GRCh38
rs768861745 Chr.11: 102350391-102350391 on Build GRCh38
rs866479373 Chr.11: 102350377-102350377 on Build GRCh38
rs376371030 Chr.11: 102350340-102350340 on Build GRCh38
rs1013822433 Chr.11: 102350444-102350444 on Build GRCh38
rs377075420 Chr.11: 102350448-102350448 on Build GRCh38
rs749151271 Chr.11: 102350361-102350361 on Build GRCh38
rs765828072 Chr.11: 102350402-102350402 on Build GRCh38
rs758975746 Chr.11: 102350416-102350416 on Build GRCh38
rs887080224 Chr.11: 102350301-102350301 on Build GRCh38
rs369921460 Chr.11: 102350369-102350369 on Build GRCh38
rs747255864 Chr.11: 102350385-102350385 on Build GRCh38
rs765773461 Chr.11: 102350322-102350322 on Build GRCh38
rs753226433 Chr.11: 102350314-102350314 on Build GRCh38
rs772282652 Chr.11: 102350383-102350383 on Build GRCh38
rs200323748 Chr.11: 102350349-102350349 on Build GRCh38
rs755833024 Chr.11: 102350431-102350431 on Build GRCh38
rs758502806 Chr.11: 102350457-102350457 on Build GRCh38
rs139349997 Chr.11: 102350379-102350379 on Build GRCh38
rs763777917 Chr.11: 102350433-102350433 on Build GRCh38
rs780311859 Chr.11: 102350467-102350467 on Build GRCh38
rs755857551 Chr.11: 102350350-102350350 on Build GRCh38
rs866704792 Chr.11: 102350428-102350428 on Build GRCh38
rs182528901 Chr.11: 102350429-102350429 on Build GRCh38
rs960568242 Chr.11: 102350333-102350333 on Build GRCh38
rs777657382 Chr.11: 102350357-102350357 on Build GRCh38
rs756595133 Chr.11: 102350487-102350487 on Build GRCh38
rs1015238864 Chr.11: 102350332-102350332 on Build GRCh38
rs202103808 Chr.11: 102350335-102350335 on Build GRCh38
rs373200978 Chr.11: 102350327-102350327 on Build GRCh38
rs764532717 Chr.11: 102350288-102350288 on Build GRCh38
rs767040556 Chr.11: 102350426-102350426 on Build GRCh38
rs768182297 Chr.11: 102350417-102350418 on Build GRCh38
rs761054060 Chr.11: 102350287-102350287 on Build GRCh38
rs563146349 Chr.11: 102350445-102350445 on Build GRCh38
rs752881236 Chr.11: 102350488-102350488 on Build GRCh38
rs910615926 Chr.11: 102350289-102350291 on Build GRCh38
rs777091998 Chr.11: 102350399-102350399 on Build GRCh38
rs142241448 Chr.11: 102350308-102350308 on Build GRCh38

View Full Product Details