Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46512557-46512782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1E2
Entrez Gene ID
90423
Gene Name
ATPase H+ transporting V1 subunit E2
Gene Aliases
ATP6E1, ATP6EL2, ATP6V1EL2, VMA4
UniGene
Hs.659656
Gene Chromosome Location
Chr.2: 46511835-46542557 on Build GRCh38
Transcripts
NM_001318063.1 XM_005264633.2 XM_005264634.2 XM_011533148.2 XM_011533149.2 XM_011533150.2 XM_011533151.2 XM_011533152.2 XM_011533153.2 XM_017005225.1 XM_017005226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3839600 c.121G>C p.E41Q Chr.2: 46512591-46512591 on Build GRCh38
7357040 c.107A>T p.E36V Chr.2: 46512605-46512605 on Build GRCh38
1752572 c.32A>G p.Q11R Chr.2: 46512680-46512680 on Build GRCh38
4392713 c.74A>T p.E25V Chr.2: 46512638-46512638 on Build GRCh38
301344 c.26A>G p.K9R Chr.2: 46512686-46512686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775137380 Chr.2: 46512656-46512664 on Build GRCh38
rs779442014 Chr.2: 46512735-46512735 on Build GRCh38
rs764530051 Chr.2: 46512592-46512592 on Build GRCh38
rs563577054 Chr.2: 46512646-46512646 on Build GRCh38
rs769209771 Chr.2: 46512672-46512672 on Build GRCh38
rs748503373 Chr.2: 46512736-46512736 on Build GRCh38
rs113101588 Chr.2: 46512581-46512581 on Build GRCh38
rs371965958 Chr.2: 46512731-46512731 on Build GRCh38
rs1005993375 Chr.2: 46512743-46512743 on Build GRCh38
rs1007883231 Chr.2: 46512723-46512723 on Build GRCh38
rs534604492 Chr.2: 46512577-46512577 on Build GRCh38
rs774789970 Chr.2: 46512674-46512674 on Build GRCh38
rs147004858 Chr.2: 46512588-46512588 on Build GRCh38
rs377582931 Chr.2: 46512596-46512596 on Build GRCh38
rs192578549 Chr.2: 46512693-46512693 on Build GRCh38
rs374785394 Chr.2: 46512625-46512625 on Build GRCh38
rs752326952 Chr.2: 46512640-46512640 on Build GRCh38
rs972052017 Chr.2: 46512580-46512580 on Build GRCh38
rs764827891 Chr.2: 46512632-46512632 on Build GRCh38
rs371491877 Chr.2: 46512664-46512664 on Build GRCh38
rs765807911 Chr.2: 46512612-46512612 on Build GRCh38
rs754606142 Chr.2: 46512622-46512622 on Build GRCh38
rs1039444387 Chr.2: 46512694-46512694 on Build GRCh38
rs201744743 Chr.2: 46512730-46512730 on Build GRCh38
rs768941974 Chr.2: 46512732-46512732 on Build GRCh38
rs756488914 Chr.2: 46512659-46512659 on Build GRCh38
rs763448899 Chr.2: 46512589-46512589 on Build GRCh38
rs762236624 Chr.2: 46512683-46512683 on Build GRCh38
rs757982839 Chr.2: 46512704-46512704 on Build GRCh38
rs781154572 Chr.2: 46512656-46512656 on Build GRCh38
rs370738793 Chr.2: 46512621-46512621 on Build GRCh38
rs751320754 Chr.2: 46512714-46512714 on Build GRCh38
rs150480219 Chr.2: 46512582-46512582 on Build GRCh38
rs533093303 Chr.2: 46512753-46512753 on Build GRCh38
rs367856598 Chr.2: 46512655-46512655 on Build GRCh38
rs746928461 Chr.2: 46512654-46512654 on Build GRCh38
rs185014020 Chr.2: 46512717-46512717 on Build GRCh38
rs763889531 Chr.2: 46512706-46512706 on Build GRCh38
rs117965097 Chr.2: 46512576-46512576 on Build GRCh38
rs541048867 Chr.2: 46512722-46512722 on Build GRCh38
rs138593929 Chr.2: 46512601-46512601 on Build GRCh38
rs764716995 Chr.2: 46512696-46512696 on Build GRCh38
rs367600039 Chr.2: 46512749-46512749 on Build GRCh38
rs777543775 Chr.2: 46512647-46512647 on Build GRCh38
rs1038631934 Chr.2: 46512684-46512684 on Build GRCh38
rs752416922 Chr.2: 46512703-46512703 on Build GRCh38
rs746718362 Chr.2: 46512689-46512689 on Build GRCh38
rs879926443 Chr.2: 46512697-46512697 on Build GRCh38
rs558643489 Chr.2: 46512725-46512725 on Build GRCh38
rs772291648 Chr.2: 46512746-46512746 on Build GRCh38
rs138145018 Chr.2: 46512691-46512691 on Build GRCh38
rs375381446 Chr.2: 46512747-46512747 on Build GRCh38
rs749507203 Chr.2: 46512671-46512671 on Build GRCh38
rs775902385 Chr.2: 46512667-46512667 on Build GRCh38
rs745923418 Chr.2: 46512658-46512658 on Build GRCh38
rs774923437 Chr.2: 46512754-46512754 on Build GRCh38

View Full Product Details