Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93003944-93004211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD3
Entrez Gene ID
89846
Gene Name
FYVE, RhoGEF and PH domain containing 3
Gene Aliases
ZFYVE5
UniGene
Hs.411081
Gene Chromosome Location
Chr.9: 92947319-93036236 on Build GRCh38
Transcripts
NM_001083536.1 NM_001286993.1 NM_033086.2 XM_017015274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5083258 c.655C>A p.L219M Chr.9: 93004112-93004112 on Build GRCh38
4501448 c.588C>T p.V196V Chr.9: 93004045-93004045 on Build GRCh38
6550223 c.635G>T p.G212V Chr.9: 93004092-93004092 on Build GRCh38
4684418 c.555C>T p.T185T Chr.9: 93004012-93004012 on Build GRCh38
1580896 c.665G>T p.R222L Chr.9: 93004122-93004122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042904384 Chr.9: 93004042-93004042 on Build GRCh38
rs376441612 Chr.9: 93004063-93004063 on Build GRCh38
rs777830355 Chr.9: 93004184-93004184 on Build GRCh38
rs762923444 Chr.9: 93004035-93004035 on Build GRCh38
rs370725925 Chr.9: 93004182-93004182 on Build GRCh38
rs895419179 Chr.9: 93004119-93004119 on Build GRCh38
rs1034501341 Chr.9: 93004012-93004012 on Build GRCh38
rs758992365 Chr.9: 93003977-93003977 on Build GRCh38
rs773396820 Chr.9: 93004029-93004029 on Build GRCh38
rs775977729 Chr.9: 93004129-93004131 on Build GRCh38
rs201073654 Chr.9: 93004054-93004054 on Build GRCh38
rs748331441 Chr.9: 93004011-93004011 on Build GRCh38
rs1022257734 Chr.9: 93004120-93004120 on Build GRCh38
rs747405516 Chr.9: 93003985-93003985 on Build GRCh38
rs371150524 Chr.9: 93004158-93004158 on Build GRCh38
rs767605391 Chr.9: 93004039-93004039 on Build GRCh38
rs1012474723 Chr.9: 93004123-93004123 on Build GRCh38
rs377157870 Chr.9: 93003978-93003978 on Build GRCh38
rs371023913 Chr.9: 93004176-93004176 on Build GRCh38
rs768959160 Chr.9: 93003986-93003986 on Build GRCh38
rs781598786 Chr.9: 93004169-93004169 on Build GRCh38
rs540510551 Chr.9: 93004162-93004162 on Build GRCh38
rs774743745 Chr.9: 93003965-93003965 on Build GRCh38
rs753545285 Chr.9: 93004111-93004111 on Build GRCh38
rs769901274 Chr.9: 93004028-93004028 on Build GRCh38
rs372428454 Chr.9: 93004107-93004107 on Build GRCh38
rs369568017 Chr.9: 93003987-93003987 on Build GRCh38
rs6479445 Chr.9: 93004189-93004189 on Build GRCh38
rs374509426 Chr.9: 93004177-93004177 on Build GRCh38
rs749988917 Chr.9: 93004136-93004136 on Build GRCh38
rs939528847 Chr.9: 93004089-93004089 on Build GRCh38
rs76713473 Chr.9: 93004149-93004149 on Build GRCh38
rs369228956 Chr.9: 93004093-93004093 on Build GRCh38
rs755336390 Chr.9: 93004166-93004166 on Build GRCh38
rs759880993 Chr.9: 93003970-93003970 on Build GRCh38
rs181243813 Chr.9: 93004112-93004112 on Build GRCh38
rs764785925 Chr.9: 93004129-93004129 on Build GRCh38
rs946726395 Chr.9: 93004013-93004013 on Build GRCh38

View Full Product Details