Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128341691-128341962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEFSEC
Entrez Gene ID
60678
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Gene Aliases
EFSEC, SELB
UniGene
Hs.477498
Gene Chromosome Location
Chr.3: 128153456-128426193 on Build GRCh38
Transcripts
NM_021937.4 XM_005247696.2 XM_011513066.2 XM_017006975.1 XM_017006976.1 XM_017006977.1 XM_017006978.1 XM_017006979.1 XM_017006980.1 XM_017006981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096315 c.1356C>T p.H452H Chr.3: 128341802-128341802 on Build GRCh38
5344340 c.1380C>G p.A460A Chr.3: 128341826-128341826 on Build GRCh38
3587098 c.1264T>G p.C422G Chr.3: 128341710-128341710 on Build GRCh38
3059495 c.1271G>A p.R424Q Chr.3: 128341717-128341717 on Build GRCh38
5698427 c.1326C>G p.C442W Chr.3: 128341772-128341772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567782092 Chr.3: 128341751-128341751 on Build GRCh38
rs370196613 Chr.3: 128341940-128341940 on Build GRCh38
rs771987158 Chr.3: 128341912-128341912 on Build GRCh38
rs374495992 Chr.3: 128341918-128341918 on Build GRCh38
rs769084139 Chr.3: 128341900-128341900 on Build GRCh38
rs970506014 Chr.3: 128341773-128341773 on Build GRCh38
rs752228050 Chr.3: 128341935-128341935 on Build GRCh38
rs758086477 Chr.3: 128341936-128341936 on Build GRCh38
rs780576987 Chr.3: 128341887-128341887 on Build GRCh38
rs1001633911 Chr.3: 128341805-128341805 on Build GRCh38
rs1021470602 Chr.3: 128341831-128341831 on Build GRCh38
rs747502656 Chr.3: 128341888-128341888 on Build GRCh38
rs775849626 Chr.3: 128341797-128341797 on Build GRCh38
rs34326479 Chr.3: 128341750-128341750 on Build GRCh38
rs750542880 Chr.3: 128341884-128341884 on Build GRCh38
rs778696433 Chr.3: 128341767-128341767 on Build GRCh38
rs913794681 Chr.3: 128341877-128341877 on Build GRCh38
rs767667061 Chr.3: 128341815-128341815 on Build GRCh38
rs754107284 Chr.3: 128341934-128341934 on Build GRCh38
rs763170278 Chr.3: 128341714-128341714 on Build GRCh38
rs776954210 Chr.3: 128341774-128341774 on Build GRCh38
rs565773364 Chr.3: 128341869-128341869 on Build GRCh38
rs754661809 Chr.3: 128341761-128341761 on Build GRCh38
rs536540678 Chr.3: 128341905-128341905 on Build GRCh38
rs755932555 Chr.3: 128341746-128341746 on Build GRCh38
rs761746184 Chr.3: 128341812-128341812 on Build GRCh38
rs747858839 Chr.3: 128341843-128341843 on Build GRCh38
rs750403689 Chr.3: 128341742-128341742 on Build GRCh38
rs766389391 Chr.3: 128341919-128341919 on Build GRCh38
rs530813952 Chr.3: 128341803-128341803 on Build GRCh38
rs746410344 Chr.3: 128341789-128341789 on Build GRCh38
rs774680437 Chr.3: 128341806-128341806 on Build GRCh38
rs764179097 Chr.3: 128341716-128341716 on Build GRCh38
rs770274023 Chr.3: 128341881-128341881 on Build GRCh38
rs200791213 Chr.3: 128341736-128341736 on Build GRCh38
rs770081501 Chr.3: 128341791-128341791 on Build GRCh38
rs552212657 Chr.3: 128341808-128341808 on Build GRCh38
rs774392055 Chr.3: 128341904-128341904 on Build GRCh38
rs920745618 Chr.3: 128341931-128341931 on Build GRCh38
rs1035465672 Chr.3: 128341896-128341896 on Build GRCh38
rs112227741 Chr.3: 128341835-128341835 on Build GRCh38
rs756132067 Chr.3: 128341824-128341824 on Build GRCh38
rs532148321 Chr.3: 128341827-128341827 on Build GRCh38
rs376378299 Chr.3: 128341930-128341930 on Build GRCh38
rs769018524 Chr.3: 128341898-128341898 on Build GRCh38
rs753877651 Chr.3: 128341926-128341926 on Build GRCh38
rs763373068 Chr.3: 128341802-128341802 on Build GRCh38
rs772141765 Chr.3: 128341863-128341868 on Build GRCh38
rs76158404 Chr.3: 128341826-128341826 on Build GRCh38
rs751583689 Chr.3: 128341717-128341717 on Build GRCh38
rs150724686 Chr.3: 128341823-128341823 on Build GRCh38
rs757810177 Chr.3: 128341847-128341847 on Build GRCh38
rs747582191 Chr.3: 128341768-128341768 on Build GRCh38
rs143447496 Chr.3: 128341769-128341769 on Build GRCh38
rs114204669 Chr.3: 128341915-128341915 on Build GRCh38
rs777345387 Chr.3: 128341859-128341859 on Build GRCh38
rs757395009 Chr.3: 128341731-128341731 on Build GRCh38

View Full Product Details