Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101539664-101539921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL26A1
Entrez Gene ID
136227
Gene Name
collagen type XXVI alpha 1
Gene Aliases
EMI6, EMID2, EMU2, SH2B
UniGene
Hs.654854
Gene Chromosome Location
Chr.7: 101362369-101559024 on Build GRCh38
Transcripts
NM_001278563.2 NM_133457.4 XM_017011743.1 XM_017011744.1 XM_017011745.1

SNP Detail

SNP ID Chromosome Location
rs377393066 Chr.7: 101539879-101539879 on Build GRCh38
rs115909926 Chr.7: 101539784-101539784 on Build GRCh38
rs538742393 Chr.7: 101539828-101539828 on Build GRCh38
rs900934476 Chr.7: 101539755-101539755 on Build GRCh38
rs762497586 Chr.7: 101539888-101539888 on Build GRCh38
rs117894148 Chr.7: 101539747-101539747 on Build GRCh38
rs900661970 Chr.7: 101539834-101539834 on Build GRCh38
rs6465817 Chr.7: 101539827-101539827 on Build GRCh38
rs758230878 Chr.7: 101539764-101539764 on Build GRCh38
rs372981039 Chr.7: 101539871-101539871 on Build GRCh38
rs929864801 Chr.7: 101539690-101539690 on Build GRCh38
rs922451327 Chr.7: 101539895-101539895 on Build GRCh38
rs550171341 Chr.7: 101539824-101539824 on Build GRCh38
rs745444238 Chr.7: 101539871-101539872 on Build GRCh38
rs1048348040 Chr.7: 101539696-101539696 on Build GRCh38
rs372116842 Chr.7: 101539804-101539804 on Build GRCh38
rs1000178023 Chr.7: 101539765-101539765 on Build GRCh38
rs756710004 Chr.7: 101539872-101539877 on Build GRCh38
rs547495355 Chr.7: 101539693-101539693 on Build GRCh38
rs942520340 Chr.7: 101539703-101539705 on Build GRCh38
rs951449724 Chr.7: 101539846-101539846 on Build GRCh38
rs565845308 Chr.7: 101539813-101539813 on Build GRCh38
rs767598146 Chr.7: 101539881-101539881 on Build GRCh38
rs548409997 Chr.7: 101539761-101539761 on Build GRCh38
rs774361643 Chr.7: 101539691-101539691 on Build GRCh38
rs979297987 Chr.7: 101539783-101539783 on Build GRCh38
rs1039435347 Chr.7: 101539703-101539703 on Build GRCh38
rs769488345 Chr.7: 101539901-101539901 on Build GRCh38
rs562881613 Chr.7: 101539812-101539812 on Build GRCh38
rs1041756004 Chr.7: 101539763-101539763 on Build GRCh38
rs184800053 Chr.7: 101539769-101539769 on Build GRCh38
rs888271342 Chr.7: 101539697-101539697 on Build GRCh38
rs1039542460 Chr.7: 101539892-101539892 on Build GRCh38
rs768564502 Chr.7: 101539854-101539854 on Build GRCh38
rs777857974 Chr.7: 101539770-101539770 on Build GRCh38
rs370315124 Chr.7: 101539897-101539897 on Build GRCh38
rs969665365 Chr.7: 101539779-101539779 on Build GRCh38
rs79591881 Chr.7: 101539759-101539759 on Build GRCh38

View Full Product Details