Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98126610-98126883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO2A
Entrez Gene ID
7464
Gene Name
coronin 2A
Gene Aliases
CLIPINB, IR10, WDR2
UniGene
Hs.113094
Gene Chromosome Location
Chr.9: 98120975-98192674 on Build GRCh38
Transcripts
NM_003389.3 NM_052820.3 XM_011518986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4394974 c.1327T>A p.S443T Chr.9: 98126668-98126668 on Build GRCh38
4976716 c.1304C>A p.A435E Chr.9: 98126691-98126691 on Build GRCh38
6837482 c.1241C>A p.P414H Chr.9: 98126754-98126754 on Build GRCh38
139794 c.1325C>T p.S442F Chr.9: 98126670-98126670 on Build GRCh38
1464347 c.1274G>T p.R425L Chr.9: 98126721-98126721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556860417 Chr.9: 98126786-98126786 on Build GRCh38
rs761409495 Chr.9: 98126730-98126730 on Build GRCh38
rs777943659 Chr.9: 98126778-98126778 on Build GRCh38
rs751327304 Chr.9: 98126864-98126864 on Build GRCh38
rs746283600 Chr.9: 98126701-98126701 on Build GRCh38
rs267602333 Chr.9: 98126667-98126667 on Build GRCh38
rs200720818 Chr.9: 98126659-98126659 on Build GRCh38
rs1017609303 Chr.9: 98126751-98126751 on Build GRCh38
rs1009394062 Chr.9: 98126750-98126750 on Build GRCh38
rs367683956 Chr.9: 98126719-98126719 on Build GRCh38
rs374959179 Chr.9: 98126757-98126757 on Build GRCh38
rs374505837 Chr.9: 98126759-98126759 on Build GRCh38
rs2231669 Chr.9: 98126863-98126863 on Build GRCh38
rs758266065 Chr.9: 98126761-98126761 on Build GRCh38
rs933709063 Chr.9: 98126715-98126715 on Build GRCh38
rs149125179 Chr.9: 98126815-98126815 on Build GRCh38
rs749409257 Chr.9: 98126781-98126781 on Build GRCh38
rs3174288 Chr.9: 98126722-98126722 on Build GRCh38
rs756277395 Chr.9: 98126656-98126656 on Build GRCh38
rs747487420 Chr.9: 98126827-98126827 on Build GRCh38
rs761354979 Chr.9: 98126847-98126847 on Build GRCh38
rs544888496 Chr.9: 98126841-98126841 on Build GRCh38
rs146552729 Chr.9: 98126721-98126721 on Build GRCh38
rs567736146 Chr.9: 98126798-98126798 on Build GRCh38
rs373733956 Chr.9: 98126670-98126670 on Build GRCh38
rs78879445 Chr.9: 98126853-98126853 on Build GRCh38
rs2231670 Chr.9: 98126835-98126835 on Build GRCh38
rs780082100 Chr.9: 98126646-98126646 on Build GRCh38
rs757521278 Chr.9: 98126783-98126783 on Build GRCh38
rs762652995 Chr.9: 98126861-98126861 on Build GRCh38
rs760096970 Chr.9: 98126720-98126720 on Build GRCh38
rs893002997 Chr.9: 98126745-98126745 on Build GRCh38
rs573279068 Chr.9: 98126740-98126740 on Build GRCh38
rs776082882 Chr.9: 98126844-98126844 on Build GRCh38
rs759039457 Chr.9: 98126853-98126853 on Build GRCh38
rs764852384 Chr.9: 98126850-98126850 on Build GRCh38
rs774816361 Chr.9: 98126692-98126692 on Build GRCh38
rs776181410 Chr.9: 98126722-98126722 on Build GRCh38
rs748158513 Chr.9: 98126650-98126650 on Build GRCh38
rs773628507 Chr.9: 98126663-98126664 on Build GRCh38
rs144845278 Chr.9: 98126642-98126642 on Build GRCh38
rs201915406 Chr.9: 98126777-98126777 on Build GRCh38
rs772434336 Chr.9: 98126714-98126714 on Build GRCh38
rs766214882 Chr.9: 98126765-98126765 on Build GRCh38
rs764668209 Chr.9: 98126864-98126864 on Build GRCh38
rs1009715793 Chr.9: 98126727-98126727 on Build GRCh38
rs111757914 Chr.9: 98126852-98126852 on Build GRCh38
rs201734917 Chr.9: 98126664-98126664 on Build GRCh38
rs751583025 Chr.9: 98126771-98126771 on Build GRCh38
rs760302166 Chr.9: 98126685-98126687 on Build GRCh38
rs766098889 Chr.9: 98126703-98126706 on Build GRCh38
rs750243854 Chr.9: 98126639-98126639 on Build GRCh38
rs375458218 Chr.9: 98126816-98126816 on Build GRCh38

View Full Product Details